Learn More
N-Ethylisopropylamine, 98%
CAS: 19961-27-4 | C5H13N | 87.17 g/mol
$60.01 - $154.82
Chemical Identifiers
| CAS | 19961-27-4 |
|---|---|
| Molecular Formula | C5H13N |
| Molecular Weight (g/mol) | 87.17 |
| MDL Number | MFCD00015105 |
| InChI Key | RIVIDPPYRINTTH-UHFFFAOYSA-N |
| Synonym | n-ethylisopropylamine, ethylisopropylamine, 2-propanamine, n-ethyl, ethyl propan-2-yl amine, diethylamine, 1-methyl, isopropylamine, n-ethyl, n-ethyl-2-propanamine, n-ethyl-n-isopropylamine, isopropylethylamine, ethyl isopropylamine |
| PubChem CID | 88318 |
| SMILES | CCNC(C)C |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAL0408903
|
Thermo Scientific Chemicals
L0408903 |
1 g |
Each for $60.01
|
|
|||||
|
AAL0408906
|
Thermo Scientific Chemicals
L0408906 |
5 g |
Each for $154.82
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 19961-27-4 | |
| 87.17 | |
| RIVIDPPYRINTTH-UHFFFAOYSA-N | |
| 88318 |
| C5H13N | |
| MFCD00015105 | |
| n-ethylisopropylamine, ethylisopropylamine, 2-propanamine, n-ethyl, ethyl propan-2-yl amine, diethylamine, 1-methyl, isopropylamine, n-ethyl, n-ethyl-2-propanamine, n-ethyl-n-isopropylamine, isopropylethylamine, ethyl isopropylamine | |
| CCNC(C)C |
Specifications
| 19961-27-4 | |
| 71°C to 73°C | |
| C5H13N | |
| MFCD00015105 | |
| UN2733 | |
| n-ethylisopropylamine, ethylisopropylamine, 2-propanamine, n-ethyl, ethyl propan-2-yl amine, diethylamine, 1-methyl, isopropylamine, n-ethyl, n-ethyl-2-propanamine, n-ethyl-n-isopropylamine, isopropylethylamine, ethyl isopropylamine | |
| CCNC(C)C | |
| 87.17 | |
| 87.17 | |
| N-Ethylisopropylamine |
| 0.72 | |
| 16°C (60°F) | |
| 1.392 | |
| 1 g | |
| 505952 | |
| RIVIDPPYRINTTH-UHFFFAOYSA-N | |
| ethyl(propan-2-yl)amine | |
| 88318 | |
| 98% |
Safety and Handling
GHS H Statement
H225-H314-H318
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.
Causes serious eye damage.
P210-P233-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
H225-H302+H312+H332-H314
DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: AMINES, FLAMMABLE, CORROSIVE, N.O.S.
EINECSNumber : 243-442-1
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only