missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N-Ethylmaleimide, 98+%, Thermo Scientific Chemicals

$127.85 - $1933.02

Chemical Identifiers

CAS 128-53-0
Molecular Formula C6H7NO2
Molecular Weight (g/mol) 125.13
InChI Key HDFGOPSGAURCEO-UHFFFAOYSA-N
Synonym n-ethylmaleimide, ethylmaleimide, 1-ethyl-1h-pyrrole-2,5-dione, maleimide, n-ethyl, 1h-pyrrole-2,5-dione, 1-ethyl, maleic acid n-ethylimide, nem, usaf b-121, unii-o3c74acm9v, maleic acid-n-ethylimide
PubChem CID 4362
ChEBI CHEBI:44485
IUPAC Name 1-ethylpyrrole-2,5-dione
SMILES CCN1C(=O)C=CC1=O
View More Specs

Products 4
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC156100050
SDS View Documents
Thermo Scientific Chemicals
156100050
5 g
Each for $127.85
 
AC156100100
SDS View Documents
Thermo Scientific Chemicals
156100100
10 g
Each for $236.91
 
AC156100500
SDS View Documents
Thermo Scientific Chemicals
156100500
50 g
Each for $643.71
 
AC156102500
SDS View Documents
Thermo Scientific Chemicals
156102500
250 g
Each for $1,933.02
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

128-53-0
125.13
n-ethylmaleimide, ethylmaleimide, 1-ethyl-1h-pyrrole-2,5-dione, maleimide, n-ethyl, 1h-pyrrole-2,5-dione, 1-ethyl, maleic acid n-ethylimide, nem, usaf b-121, unii-o3c74acm9v, maleic acid-n-ethylimide
CHEBI:44485
CCN1C(=O)C=CC1=O
C6H7NO2
HDFGOPSGAURCEO-UHFFFAOYSA-N
4362
1-ethylpyrrole-2,5-dione

Specifications

128-53-0
White
73°C
99% min. (GC)
C6H7NO2
21,IV,4629
n-ethylmaleimide, ethylmaleimide, 1-ethyl-1h-pyrrole-2,5-dione, maleimide, n-ethyl, 1h-pyrrole-2,5-dione, 1-ethyl, maleic acid n-ethylimide, nem, usaf b-121, unii-o3c74acm9v, maleic acid-n-ethylimide
HDFGOPSGAURCEO-UHFFFAOYSA-N
1-ethylpyrrole-2,5-dione
4362
125.13
Crystalline Powder
41.0°C to 45.0°C
210.0°C
Authentic
Glass bottle
5 g
15,3877
Solubility in water: soluble. Other solubilities: 1115g/L toluene (20°C)
CCN1C(=O)C=CC1=O
125.13
CHEBI:44485
99+%
N-Ethylmaleimide
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Fatal if swallowed.
Toxic in contact with skin.
Causes severe skin burns and eye damage.
May cause an allergic skin reaction.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre

GHS Signal Word: Danger

EINECSNumber : 204-892-4

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only