missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N-Ethylmethylamine, 98+%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals L0437206

 View more versions of this product

Catalog No. AAL0437206


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

N-Ethylmethylamine
0.688
<−34°C (−29°F)
C3H9N
MFCD00009030
UN2733
Air sensitive; Hygroscopic
LIWAQLJGPBVORC-UHFFFAOYSA-N
N-methylethanamine
12219
≥98%
624-78-2
33°C to 34°C
Amine-like
1.374
5 g
1730786
n-ethylmethylamine, methylethylamine, ethanamine, n-methyl, n-methylethylamine, ethyl methyl amine, ethylmethylamine, nmea, n-methyl-n-ethylamine, ethylamine, n-methyl, ethyl-methyl-amine
CCNC
59.11
59.11

Chemical Identifiers

624-78-2
59.11
LIWAQLJGPBVORC-UHFFFAOYSA-N
12219
CCNC
C3H9N
MFCD00009030
n-ethylmethylamine, methylethylamine, ethanamine, n-methyl, n-methylethylamine, ethyl methyl amine, ethylmethylamine, nmea, n-methyl-n-ethylamine, ethylamine, n-methyl, ethyl-methyl-amine
N-methylethanamine
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H224-H314-H318
Extremely flammable liquid and vapor.
Causes severe skin burns and eye damage.
Causes serious eye damage.

P210-P233-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c

H225-H302+H312+H332-H314

DOTInformation : Transport Hazard Class: 3; Packing Group: I; Proper Shipping Name: AMINES, FLAMMABLE, CORROSIVE, N.O.S.

EINECSNumber : 210-862-1

TSCA : Yes

Recommended Storage : Keep cold

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only