missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Ergodyne™ N-Ferno™ 6471 Thermal Bibs/Overalls
Insulated bibs/overalls provide the ultimate in durability, weather protection, and quality outerwear
$339.30 - $339.30
Specifications
Color | Black |
---|---|
Material | Nylon (500D) |
For Use With (Application) | Roof Bolting, Mining, Oil/Gas, Demolition, Heavy Construction, Ironwork, Mechanics |
Catalog Number | Mfr. No. | Size | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Size | Price | Quantity | |||||
19-910-194
|
Ergodyne™
41212 |
Small |
Each for $339.30
|
|
|||||
19-910-195
|
Ergodyne™
41213 |
Medium |
Each for $339.30
|
|
|||||
19-910-196
|
Ergodyne™
41214 |
Large |
Each for $339.30
|
|
|||||
19-910-197
|
Ergodyne™
41215 |
X-Large |
Each for $339.30
|
|
|||||
19-910-198
|
Ergodyne™
41216 |
2X-Large |
Each for $339.30
|
|
|||||
19-910-199
|
Ergodyne™
41217 |
3X-Large |
Each for $339.30
|
|
|||||
Description
Features removable knee/shin pads and 1600D nylon reinforcement in heavy wear areasSpecifications
Black | |
Nylon (500D) | |
Roof Bolting, Mining, Oil/Gas, Demolition, Heavy Construction, Ironwork, Mechanics |