missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
N-Fmoc-3-(2-naphthyl)-D-alanine, 95%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H5197406
Description
N-Fmoc-3-(2-naphthyl)-D-alanine is an Fmoc-protected alanine derivative and an intermediate used in the preparation of phosphoserine-containing tetrapeptides with hydrophobic N-terminal acyl groups as inhibitors of the BRCA1 protein.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsN-Fmoc-3-(2-naphthyl)-D-alanine is an Fmoc-protected alanine derivative and an intermediate used in the preparation of phosphoserine-containing tetrapeptides with hydrophobic N-terminal acyl groups as inhibitors of the BRCA1 protein.
Solubility
Slightly soluble in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications
N-Fmoc-3-(2-naphthyl)-D-alanine | |
C28H23NO4 | |
5 g | |
fmoc-d-2-nal-oh, fmoc-3-2-naphthyl-d-alanine, r-2-9h-fluoren-9-yl methoxy carbonyl amino-3-naphthalen-2-yl propanoic acid, fmoc-d-3-2-naphthyl-alanine, n-fmoc-3-2-naphthyl-d-alanine, fmoc-d-2-naphthylalanine, s-n-fmoc-2-naphthylalanine, fmoc-beta-2-naphthyl-d-alanine, 2r-2-9h-fluoren-9-ylmethoxy carbonyl amino-3-naphthalen-2-yl propanoic acid, pubchem11948 | |
JYUTZJVERLGMQZ-AREMUKBSSA-N | |
(2R)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-naphthalen-2-ylpropanoic acid | |
7130590 | |
95% |
138774-94-4 | |
MFCD00077044 | |
7864489 | |
Slightly soluble in water. | |
OC(=O)[C@@H](CC1=CC2=CC=CC=C2C=C1)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12 | |
437.50 | |
437.49 |
Chemical Identifiers
138774-94-4 | |
437.50 | |
JYUTZJVERLGMQZ-AREMUKBSSA-N | |
7130590 | |
OC(=O)[C@@H](CC1=CC2=CC=CC=C2C=C1)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12 |
C28H23NO4 | |
MFCD00077044 | |
fmoc-d-2-nal-oh, fmoc-3-2-naphthyl-d-alanine, r-2-9h-fluoren-9-yl methoxy carbonyl amino-3-naphthalen-2-yl propanoic acid, fmoc-d-3-2-naphthyl-alanine, n-fmoc-3-2-naphthyl-d-alanine, fmoc-d-2-naphthylalanine, s-n-fmoc-2-naphthylalanine, fmoc-beta-2-naphthyl-d-alanine, 2r-2-9h-fluoren-9-ylmethoxy carbonyl amino-3-naphthalen-2-yl propanoic acid, pubchem11948 | |
(2R)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-naphthalen-2-ylpropanoic acid |
Safety and Handling
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only