missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Fmoc-3-(3-thienyl)-L-alanine, 95%
CAS: 186320-06-9 | C22H19NO4S | 393.457 g/mol
$141.59 - $480.16
Chemical Identifiers
| CAS | 186320-06-9 |
|---|---|
| Molecular Formula | C22H19NO4S |
| Molecular Weight (g/mol) | 393.457 |
| MDL Number | MFCD00151916 |
| InChI Key | LSBZJMRHROCYGY-FQEVSTJZSA-N |
| Synonym | fmoc-l-3-thienylalanine, fmoc-3-ala 3-thienyl-oh, s-2-9h-fluoren-9-yl methoxy carbonyl amino-3-thiophen-3-yl propanoic acid, fmoc-beta-3-thienyl-ala-oh, fmoc-3-3-thienyl-l-alanine, 2s-2-9h-fluoren-9-ylmethoxy carbonyl amino-3-thiophen-3-yl propanoic acid, fmoc-d-3-3-thienyl-alanine, fmoc-l-3-3-thienyl alanine, n-fmoc-3-4-thienyl-l-alanine, fmoc-3 thg-oh |
| PubChem CID | 2734455 |
| IUPAC Name | (2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-thiophen-3-ylpropanoic acid |
| SMILES | C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)NC(CC4=CSC=C4)C(=O)O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH52167MD
|
Thermo Scientific Chemicals
H52167MD |
250 mg |
Each for $141.59
|
|
|||||
|
AAH5216703
|
Thermo Scientific Chemicals
H5216703 |
1 g |
Each for $480.16
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 186320-06-9 | |
| 393.457 | |
| LSBZJMRHROCYGY-FQEVSTJZSA-N | |
| 2734455 | |
| C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)NC(CC4=CSC=C4)C(=O)O |
| C22H19NO4S | |
| MFCD00151916 | |
| fmoc-l-3-thienylalanine, fmoc-3-ala 3-thienyl-oh, s-2-9h-fluoren-9-yl methoxy carbonyl amino-3-thiophen-3-yl propanoic acid, fmoc-beta-3-thienyl-ala-oh, fmoc-3-3-thienyl-l-alanine, 2s-2-9h-fluoren-9-ylmethoxy carbonyl amino-3-thiophen-3-yl propanoic acid, fmoc-d-3-3-thienyl-alanine, fmoc-l-3-3-thienyl alanine, n-fmoc-3-4-thienyl-l-alanine, fmoc-3 thg-oh | |
| (2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-thiophen-3-ylpropanoic acid |
Specifications
| 186320-06-9 | |
| MFCD00151916 | |
| 9512976 | |
| LSBZJMRHROCYGY-FQEVSTJZSA-N | |
| (2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-thiophen-3-ylpropanoic acid | |
| 2734455 | |
| 95% |
| C22H19NO4S | |
| 250 mg | |
| fmoc-l-3-thienylalanine, fmoc-3-ala 3-thienyl-oh, s-2-9h-fluoren-9-yl methoxy carbonyl amino-3-thiophen-3-yl propanoic acid, fmoc-beta-3-thienyl-ala-oh, fmoc-3-3-thienyl-l-alanine, 2s-2-9h-fluoren-9-ylmethoxy carbonyl amino-3-thiophen-3-yl propanoic acid, fmoc-d-3-3-thienyl-alanine, fmoc-l-3-3-thienyl alanine, n-fmoc-3-4-thienyl-l-alanine, fmoc-3 thg-oh | |
| C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)NC(CC4=CSC=C4)C(=O)O | |
| 393.457 | |
| 393.46 | |
| N-Fmoc-3-(3-thienyl)-L-alanine |
Safety and Handling
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only