missing translation for 'onlineSavingsMsg'
Learn More

N-Fmoc-4-chloro-L-phenylalanine, 98%

CAS: 175453-08-4 | C24H20ClNO4 | 421.877 g/mol

$175.29 - $584.32

Chemical Identifiers

CAS 175453-08-4
Molecular Formula C24H20ClNO4
Molecular Weight (g/mol) 421.877
MDL Number MFCD00080281
InChI Key CQPNKLNINBUUOM-QFIPXVFZSA-N
Synonym fmoc-4-chloro-l-phenylalanine, s-n-fmoc-4-chlorophenylalanine, fmoc-phe 4-cl-oh, fmoc-l-4-chlorophenylalanine, fmoc-l-phe 4-cl-oh, fmoc-4-chloro-phe-oh, fmoc-l-4-chlorophe, 2s-3-4-chlorophenyl-2-9h-fluoren-9-ylmethoxycarbonylamino propanoic acid, s-2-9h-fluoren-9-yl methoxy carbonyl amino-3-4-chlorophenyl propanoic acid, phenylalanine, 4-chloro-n-9h-fluoren-9-ylmethoxy carbonyl
PubChem CID 2734471
IUPAC Name (2S)-3-(4-chlorophenyl)-2-(9H-fluoren-9-ylmethoxycarbonylamino)propanoic acid
SMILES C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)NC(CC4=CC=C(C=C4)Cl)C(=O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6344803
View Documents
Thermo Scientific Chemicals
H6344803
1 g
Each for $175.29
Only null left
 
AAH6344806
View Documents
Thermo Scientific Chemicals
H6344806
5 g
Each for $584.32
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

175453-08-4
421.877
CQPNKLNINBUUOM-QFIPXVFZSA-N
2734471
C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)NC(CC4=CC=C(C=C4)Cl)C(=O)O
C24H20ClNO4
MFCD00080281
fmoc-4-chloro-l-phenylalanine, s-n-fmoc-4-chlorophenylalanine, fmoc-phe 4-cl-oh, fmoc-l-4-chlorophenylalanine, fmoc-l-phe 4-cl-oh, fmoc-4-chloro-phe-oh, fmoc-l-4-chlorophe, 2s-3-4-chlorophenyl-2-9h-fluoren-9-ylmethoxycarbonylamino propanoic acid, s-2-9h-fluoren-9-yl methoxy carbonyl amino-3-4-chlorophenyl propanoic acid, phenylalanine, 4-chloro-n-9h-fluoren-9-ylmethoxy carbonyl
(2S)-3-(4-chlorophenyl)-2-(9H-fluoren-9-ylmethoxycarbonylamino)propanoic acid

Specifications

175453-08-4
MFCD00080281
fmoc-4-chloro-l-phenylalanine, s-n-fmoc-4-chlorophenylalanine, fmoc-phe 4-cl-oh, fmoc-l-4-chlorophenylalanine, fmoc-l-phe 4-cl-oh, fmoc-4-chloro-phe-oh, fmoc-l-4-chlorophe, 2s-3-4-chlorophenyl-2-9h-fluoren-9-ylmethoxycarbonylamino propanoic acid, s-2-9h-fluoren-9-yl methoxy carbonyl amino-3-4-chlorophenyl propanoic acid, phenylalanine, 4-chloro-n-9h-fluoren-9-ylmethoxy carbonyl
C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)NC(CC4=CC=C(C=C4)Cl)C(=O)O
1 g
2734471
98%
N-Fmoc-4-chloro-L-phenylalanine
C24H20ClNO4
9437838
CQPNKLNINBUUOM-QFIPXVFZSA-N
(2S)-3-(4-chlorophenyl)-2-(9H-fluoren-9-ylmethoxycarbonylamino)propanoic acid
421.877
421.87
Solid
Videos
Safety and Handling

Safety and Handling

TSCA : No

storageNote1 : 2° to 8°C

Recommended Storage : Keep cold

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only