missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N-Fmoc-glycine, 98%, Thermo Scientific Chemicals

 

$50.59 - $172.58

Chemical Identifiers

CAS 29022-11-5
Molecular Formula C17H15NO4
Molecular Weight (g/mol) 297.31
MDL Number MFCD00037140
InChI Key NDKDFTQNXLHCGO-UHFFFAOYSA-N
Synonym fmoc-gly-oh, fmoc-glycine, n-9h-fluoren-9-ylmethoxy carbonyl glycine, n-fmoc-glycine, fmoc-dl-glycine, n-9-fmoc-l-glycine, n-alpha-fmoc-glycine, fmoc-gly, n-9-fluorenylmethoxycarbonyl glycine
PubChem CID 93124
IUPAC Name 2-(9H-fluoren-9-ylmethoxycarbonylamino)acetic acid
SMILES OC(=O)CNC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAB2105003
View Documents
Thermo Scientific Chemicals
B2105003
1 g
Each for $50.59
 
AAB2105006
View Documents
Thermo Scientific Chemicals
B2105006
5 g
Each for $172.58
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

 
Specifications

Chemical Identifiers

29022-11-5
297.31
NDKDFTQNXLHCGO-UHFFFAOYSA-N
93124
OC(=O)CNC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12
C17H15NO4
MFCD00037140
fmoc-gly-oh, fmoc-glycine, n-9h-fluoren-9-ylmethoxy carbonyl glycine, n-fmoc-glycine, fmoc-dl-glycine, n-9-fmoc-l-glycine, n-alpha-fmoc-glycine, fmoc-gly, n-9-fluorenylmethoxycarbonyl glycine
2-(9H-fluoren-9-ylmethoxycarbonylamino)acetic acid

Specifications

29022-11-5
C17H15NO4
1 g
fmoc-gly-oh, fmoc-glycine, n-9h-fluoren-9-ylmethoxy carbonyl glycine, n-fmoc-glycine, fmoc-dl-glycine, n-9-fmoc-l-glycine, n-alpha-fmoc-glycine, fmoc-gly, n-9-fluorenylmethoxycarbonyl glycine
OC(=O)CNC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12
297.31
297.32
N-Fmoc-glycine
172°C to 175°C
MFCD00037140
2163967
NDKDFTQNXLHCGO-UHFFFAOYSA-N
2-(9H-fluoren-9-ylmethoxycarbonylamino)acetic acid
93124
98%
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 249-373-3

TSCA : No

Recommended Storage : Keep cold

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only