missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
N-Fmoc-L-aspartic acid 1-tert-butyl ester, 95%, Thermo Scientific Chemicals
$63.92 - $202.71
Chemical Identifiers
CAS | 129460-09-9 |
---|---|
Molecular Formula | C23H25NO6 |
Molecular Weight (g/mol) | 411.45 |
MDL Number | MFCD00065631 |
InChI Key | VZXQYACYLGRQJU-UHFFFAOYNA-N |
Synonym | fmoc-asp-otbu, fmoc-asp-obut, s-3-9h-fluoren-9-yl methoxy carbonyl amino-4-tert-butoxy-4-oxobutanoic acid, l-fmoc-aspartic acid alpha-t-butyl ester, fmoc-asp oh-otbu, n-fmoc-l-aspartic acid 1-tert-butyl ester, 1-tert-butyl n-9h-fluoren-9-ylmethoxy carbonyl-l-aspartate, 3s-4-tert-butoxy-3-9h-fluoren-9-ylmethoxy carbonyl amino-4-oxobutanoic acid, 1-tert-butyl n-fmoc-l-aspartate, fmoc asp-otbu |
PubChem CID | 7017910 |
SMILES | CC(C)(C)OC(=O)C(CC(O)=O)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAH6235503
|
Thermo Scientific Chemicals
H6235503 |
1 g |
Each for $63.92
|
|
|||||
AAH6235506
|
Thermo Scientific Chemicals
H6235506 |
5 g |
Each for $202.71
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
129460-09-9 | |
411.45 | |
VZXQYACYLGRQJU-UHFFFAOYNA-N | |
7017910 |
C23H25NO6 | |
MFCD00065631 | |
fmoc-asp-otbu, fmoc-asp-obut, s-3-9h-fluoren-9-yl methoxy carbonyl amino-4-tert-butoxy-4-oxobutanoic acid, l-fmoc-aspartic acid alpha-t-butyl ester, fmoc-asp oh-otbu, n-fmoc-l-aspartic acid 1-tert-butyl ester, 1-tert-butyl n-9h-fluoren-9-ylmethoxy carbonyl-l-aspartate, 3s-4-tert-butoxy-3-9h-fluoren-9-ylmethoxy carbonyl amino-4-oxobutanoic acid, 1-tert-butyl n-fmoc-l-aspartate, fmoc asp-otbu | |
CC(C)(C)OC(=O)C(CC(O)=O)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12 |
Specifications
129460-09-9 | |
MFCD00065631 | |
3658045 | |
VZXQYACYLGRQJU-UHFFFAOYNA-N | |
(3S)-3-(9H-fluoren-9-ylmethoxycarbonylamino)-4-[(2-methylpropan-2-yl)oxy]-4-oxobutanoic acid | |
7017910 | |
95% |
C23H25NO6 | |
1 g | |
fmoc-asp-otbu, fmoc-asp-obut, s-3-9h-fluoren-9-yl methoxy carbonyl amino-4-tert-butoxy-4-oxobutanoic acid, l-fmoc-aspartic acid alpha-t-butyl ester, fmoc-asp oh-otbu, n-fmoc-l-aspartic acid 1-tert-butyl ester, 1-tert-butyl n-9h-fluoren-9-ylmethoxy carbonyl-l-aspartate, 3s-4-tert-butoxy-3-9h-fluoren-9-ylmethoxy carbonyl amino-4-oxobutanoic acid, 1-tert-butyl n-fmoc-l-aspartate, fmoc asp-otbu | |
CC(C)(C)OC(=O)C(CC(O)=O)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12 | |
411.45 | |
411.45 | |
N-Fmoc-L-aspartic acid 1-tert-butyl ester |
Safety and Handling
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only