missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
N-Fmoc-L-beta-homophenylalanine, 95%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H5205203
Description
N-Fmoc-L-beta-homophenylalanine is used for producing high quality peptides.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsN-Fmoc-L-beta-homophenylalanine is used for producing high quality peptides.
Solubility
Slightly soluble in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications
N-Fmoc-L-beta-homophenylalanine | |
C25H23NO4 | |
1 g | |
fmoc-l-beta-homophenylalanine, fmoc-s-3-amino-4-phenylbutyric acid, fmoc-beta-hophe-oh, fmoc-beta-hophe-oh, fmoc-beta-homophe-oh, 3s-3-9h-fluoren-9-ylmethoxy carbonyl amino-4-phenylbutanoic acid, fmoc-, fmoc-, a-hophe-oh, n-fmoc-l-beta-homophenylalanine, n-fmoc-l-beta-homophenylalanine | |
DQNUGHJJKNFCND-SFHVURJKSA-N | |
(3S)-3-(9H-fluoren-9-ylmethoxycarbonylamino)-4-phenylbutanoic acid | |
2761539 | |
95% |
193954-28-8 | |
MFCD01863055 | |
7726557 | |
Slightly soluble in water. | |
C1=CC=C(C=C1)CC(CC(=O)O)NC(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24 | |
401.462 | |
401.46 |
Chemical Identifiers
193954-28-8 | |
401.462 | |
DQNUGHJJKNFCND-SFHVURJKSA-N | |
2761539 | |
C1=CC=C(C=C1)CC(CC(=O)O)NC(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24 |
C25H23NO4 | |
MFCD01863055 | |
fmoc-l-beta-homophenylalanine, fmoc-s-3-amino-4-phenylbutyric acid, fmoc-beta-hophe-oh, fmoc-beta-hophe-oh, fmoc-beta-homophe-oh, 3s-3-9h-fluoren-9-ylmethoxy carbonyl amino-4-phenylbutanoic acid, fmoc-, fmoc-, a-hophe-oh, n-fmoc-l-beta-homophenylalanine, n-fmoc-l-beta-homophenylalanine | |
(3S)-3-(9H-fluoren-9-ylmethoxycarbonylamino)-4-phenylbutanoic acid |
Safety and Handling
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only