missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
N-Fmoc-L-glutamic acid 1-allyl ester, 98%, Thermo Scientific Chemicals
$51.63 - $461.63
Chemical Identifiers
CAS | 144120-54-7 |
---|---|
Molecular Formula | C23H23NO6 |
Molecular Weight (g/mol) | 409.44 |
MDL Number | MFCD00467718 |
InChI Key | ORKKMGRINLTBPC-UHFFFAOYNA-N |
Synonym | fmoc-glu-oall, n-9h-fluorene-9-ylmethoxycarbonyl-l-glutamic acid 1-allyl ester, s-4-9h-fluoren-9-yl methoxy carbonyl amino-5-allyloxy-5-oxopentanoic acid, s-4-9h-fluoren-9-yl methoxy carbonyl amino-5-allyloxy-5-oxopentanoic acid, a-allyl ester, fmoc-glu-allyl ester, fmoc-glu-allyl ester, 1-allyl n-fmoc-l-glutamate, fmoc-glu-oall hplc, 1-allyl n-9h-fluoren-9-ylmethoxy carbonyl-l-glutamate, 1-allyl n-9h-fluoren-9-ylmethoxy carbonyl-l-glutamate |
PubChem CID | 7020606 |
IUPAC Name | 4-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-5-oxo-5-(prop-2-en-1-yloxy)pentanoic acid |
SMILES | OC(=O)CCC(NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(=O)OCC=C |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAH66448MD
|
Thermo Scientific Chemicals
H66448MD |
250 mg |
Each for $51.63
|
|
|||||
AAH6644803
|
Thermo Scientific Chemicals
H6644803 |
1 g |
Each for $135.26
|
|
|||||
AAH6644806
|
Thermo Scientific Chemicals
H6644806 |
5 g |
Each for $461.63
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
144120-54-7 | |
409.44 | |
ORKKMGRINLTBPC-UHFFFAOYNA-N | |
7020606 | |
OC(=O)CCC(NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(=O)OCC=C |
C23H23NO6 | |
MFCD00467718 | |
fmoc-glu-oall, n-9h-fluorene-9-ylmethoxycarbonyl-l-glutamic acid 1-allyl ester, s-4-9h-fluoren-9-yl methoxy carbonyl amino-5-allyloxy-5-oxopentanoic acid, s-4-9h-fluoren-9-yl methoxy carbonyl amino-5-allyloxy-5-oxopentanoic acid, a-allyl ester, fmoc-glu-allyl ester, fmoc-glu-allyl ester, 1-allyl n-fmoc-l-glutamate, fmoc-glu-oall hplc, 1-allyl n-9h-fluoren-9-ylmethoxy carbonyl-l-glutamate, 1-allyl n-9h-fluoren-9-ylmethoxy carbonyl-l-glutamate | |
4-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-5-oxo-5-(prop-2-en-1-yloxy)pentanoic acid |
Specifications
118°C to 122°C | |
Brown | |
MFCD00467718 | |
fmoc-glu-oall, fmoc-l-glutamic acid 1-allyl ester, fmoc-l-glutamic acid 1-allyl ester, s-4-9h-fluoren-9-yl methoxy carbonyl amino-5-allyloxy-5-oxopentanoic acid, a-allyl ester, a-allyl ester, fmoc-glu-allyl ester, n-fmoc-l-glutamic acid 1-allyl ester, n-fmoc-l-glutamic acid 1-allyl ester, 1-allyl n-9h-fluoren-9-ylmethoxy carbonyl-l-glutamate, 1-allyl n-9h-fluoren-9-ylmethoxy carbonyl-l-glutamate | |
OC(=O)CCC(NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(=O)OCC=C | |
409.44 | |
409.44 | |
250 mg | |
N-Fmoc-L-glutamic acid 1-allyl ester |
144120-54-7 | |
C23H23NO6 | |
5460531 | |
ORKKMGRINLTBPC-UHFFFAOYNA-N | |
4-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-5-oxo-5-(prop-2-en-1-yloxy)pentanoic acid | |
7020606 | |
98% | |
Powder |
Safety and Handling
TSCA : No
storageNote1 : 2° to 8°C
Recommended Storage : Keep cold
RUO – Research Use Only