missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
N-Fmoc-L-glutamic acid 5-benzyl ester, 95%, Thermo Scientific™
$115.89 - $115.89
Chemical Identifiers
CAS | 123639-61-2 |
---|---|
Molecular Formula | C27H25NO6 |
Molecular Weight (g/mol) | 459.498 |
MDL Number | MFCD00065642 |
InChI Key | HJJURMMMGPQIQP-DEOSSOPVSA-N |
Synonym | fmoc-glu obzl-oh, fmoc-l-glutamic acid-gamma-benzyl ester, s-2-9h-fluoren-9-yl methoxy carbonyl amino-5-benzyloxy-5-oxopentanoic acid, fmoc-l-glutamic acid 5-benzyl ester, 2s-5-benzyloxy-2-9h-fluoren-9-ylmethoxy carbonyl amino-5-oxopentanoic acid, pubchem10013, fmoc-glu obzl-oh hplc, n-9-fluorenylmethoxycarbonyl-l-glutamic acid-gamma-benzyl ester, n-alpha-9-fluorenylmethyloxycarbonyl-l-glutamic-acid-gamma-benzyl ester, 2s-2-fluoren-9-ylmethoxy carbonylamino-4-benzyloxycarbonyl butanoic acid |
PubChem CID | 13966932 |
IUPAC Name | (2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-5-oxo-5-phenylmethoxypentanoic acid |
SMILES | C1=CC=C(C=C1)COC(=O)CCC(C(=O)O)NC(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAH6278306
|
Thermo Scientific Chemicals
H6278306 |
5 g |
Each for $115.89
|
|
|||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
123639-61-2 | |
459.498 | |
HJJURMMMGPQIQP-DEOSSOPVSA-N | |
13966932 | |
C1=CC=C(C=C1)COC(=O)CCC(C(=O)O)NC(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24 |
C27H25NO6 | |
MFCD00065642 | |
fmoc-glu obzl-oh, fmoc-l-glutamic acid-gamma-benzyl ester, s-2-9h-fluoren-9-yl methoxy carbonyl amino-5-benzyloxy-5-oxopentanoic acid, fmoc-l-glutamic acid 5-benzyl ester, 2s-5-benzyloxy-2-9h-fluoren-9-ylmethoxy carbonyl amino-5-oxopentanoic acid, pubchem10013, fmoc-glu obzl-oh hplc, n-9-fluorenylmethoxycarbonyl-l-glutamic acid-gamma-benzyl ester, n-alpha-9-fluorenylmethyloxycarbonyl-l-glutamic-acid-gamma-benzyl ester, 2s-2-fluoren-9-ylmethoxy carbonylamino-4-benzyloxycarbonyl butanoic acid | |
(2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-5-oxo-5-phenylmethoxypentanoic acid |
Specifications
123639-61-2 | |
C27H25NO6 | |
5 g | |
fmoc-glu obzl-oh, fmoc-l-glutamic acid-gamma-benzyl ester, s-2-9h-fluoren-9-yl methoxy carbonyl amino-5-benzyloxy-5-oxopentanoic acid, fmoc-l-glutamic acid 5-benzyl ester, 2s-5-benzyloxy-2-9h-fluoren-9-ylmethoxy carbonyl amino-5-oxopentanoic acid, pubchem10013, fmoc-glu obzl-oh hplc, n-9-fluorenylmethoxycarbonyl-l-glutamic acid-gamma-benzyl ester, n-alpha-9-fluorenylmethyloxycarbonyl-l-glutamic-acid-gamma-benzyl ester, 2s-2-fluoren-9-ylmethoxy carbonylamino-4-benzyloxycarbonyl butanoic acid | |
+12° (c=1 in Chloroform) | |
(2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-5-oxo-5-phenylmethoxypentanoic acid | |
13966932 | |
95% |
(decomposition) | |
MFCD00065642 | |
3576722 | |
HJJURMMMGPQIQP-DEOSSOPVSA-N | |
C1=CC=C(C=C1)COC(=O)CCC(C(=O)O)NC(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24 | |
459.498 | |
459.49 | |
N-Fmoc-L-glutamic acid 5-benzyl ester |
Safety and Handling
TSCA : No
Recommended Storage : Keep cold