missing translation for 'onlineSavingsMsg'
Learn More

N-Fmoc-L-leucine, 98%

CAS: 35661-60-0 | C21H23NO4 | 353.42 g/mol

$83.14 - $229.49

Chemical Identifiers

CAS 35661-60-0
Molecular Formula C21H23NO4
Molecular Weight (g/mol) 353.42
MDL Number MFCD00037133
InChI Key CBPJQFCAFFNICX-TWYLJJHKNA-N
Synonym fmoc-l-leucine, fmoc-leu-oh, fmoc-l-leu-oh, n-fmoc-l-leucine, n-9-fluorenylmethoxycarbonyl-l-leucine, fmoc-leucine, n-9h-fluoren-9-ylmethoxy carbonyl-l-leucine, unii-kf18f70uf3, fmoc-leu, n-alpha-fmoc-l-leucine
PubChem CID 1549133
IUPAC Name (2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-4-methylpentanoic acid
SMILES CC(C)C[C@H](NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAB2104006
View Documents
Thermo Scientific Chemicals
B2104006
5 g
Each for $83.14
Only null left
 
AAB2104014
View Documents
Thermo Scientific Chemicals
B2104014
25 g
Each for $229.49
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

35661-60-0
353.42
CBPJQFCAFFNICX-TWYLJJHKNA-N
1549133
CC(C)C[C@H](NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O
C21H23NO4
MFCD00037133
fmoc-l-leucine, fmoc-leu-oh, fmoc-l-leu-oh, n-fmoc-l-leucine, n-9-fluorenylmethoxycarbonyl-l-leucine, fmoc-leucine, n-9h-fluoren-9-ylmethoxy carbonyl-l-leucine, unii-kf18f70uf3, fmoc-leu, n-alpha-fmoc-l-leucine
(2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-4-methylpentanoic acid

Specifications

35661-60-0
C21H23NO4
5 g
fmoc-l-leucine, fmoc-leu-oh, fmoc-l-leu-oh, n-fmoc-l-leucine, n-9-fluorenylmethoxycarbonyl-l-leucine, fmoc-leucine, n-9h-fluoren-9-ylmethoxy carbonyl-l-leucine, unii-kf18f70uf3, fmoc-leu, n-alpha-fmoc-l-leucine
−25° (c=1 in DMF)
(2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-4-methylpentanoic acid
1549133
98%
152°C to 155°C
MFCD00037133
2178254
CBPJQFCAFFNICX-TWYLJJHKNA-N
CC(C)C[C@H](NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O
353.42
353.42
N-Fmoc-L-leucine
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 252-662-7

TSCA : No

Recommended Storage : Keep cold

SDS
Documents

Documents

RUO – Research Use Only