missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N-Fmoc-L-propargylglycine, 95%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals H5217106

 View more versions of this product

Catalog No. AAH5217106


Explore available promotions

Description

Description

Fmoc-propargyl-Gly-OH is an Fmoc protected glycine derivative useful for solid phase peptide synthesis techniques. This compound could be useful as an unusual amino acid analog to aid in the deconvolution of protein structure and function. N-Fmoc-L-propargylglycine is a non-natural amino acids used for the triazole cross-link.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Fmoc-propargyl-Gly-OH is an Fmoc protected glycine derivative useful for solid phase peptide synthesis techniques. This compound could be useful as an unusual amino acid analog to aid in the deconvolution of protein structure and function. N-Fmoc-L-propargylglycine is a non-natural amino acids used for the triazole cross-link.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents. Store at 4°C.
Specifications

Specifications

N-Fmoc-L-propargylglycine
C20H17NO4
5 g
fmoc-l-propargylglycine, fmoc-pra-oh, fmoc-propargyl-gly-oh, s-2-fmoc-amino-4-pentynoic acid, s-n-fmoc-2-2'-propynyl glycine, 2s-2-9h-fluoren-9-ylmethoxycarbonylamino pent-4-ynoic acid, 2s-2-9h-fluoren-9-ylmethoxy carbonyl amino pent-4-ynoic acid, s-2-9h-fluoren-9-yl methoxy carbonyl amino pent-4-ynoic acid, 4-pentynoic acid, 2-9h-fluoren-9-ylmethoxy carbonyl amino-, 2s, 4-pentynoicacid, 2-9h-fluoren-9-ylmethoxy carbonyl amino-, 2s
C#CCC(C(=O)O)NC(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13
335.359
335.36
198561-07-8
MFCD01075095
9151699
DJGMNCKHNMRKFM-SFHVURJKSA-N
(2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)pent-4-ynoic acid
2734461
95%

Chemical Identifiers

198561-07-8
335.359
DJGMNCKHNMRKFM-SFHVURJKSA-N
2734461
C#CCC(C(=O)O)NC(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13
C20H17NO4
MFCD01075095
fmoc-l-propargylglycine, fmoc-pra-oh, fmoc-propargyl-gly-oh, s-2-fmoc-amino-4-pentynoic acid, s-n-fmoc-2-2'-propynyl glycine, 2s-2-9h-fluoren-9-ylmethoxycarbonylamino pent-4-ynoic acid, 2s-2-9h-fluoren-9-ylmethoxy carbonyl amino pent-4-ynoic acid, s-2-9h-fluoren-9-yl methoxy carbonyl amino pent-4-ynoic acid, 4-pentynoic acid, 2-9h-fluoren-9-ylmethoxy carbonyl amino-, 2s, 4-pentynoicacid, 2-9h-fluoren-9-ylmethoxy carbonyl amino-, 2s
(2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)pent-4-ynoic acid
Videos
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only