missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N-Fmoc-N-methyl-L-norleucine, 96%, Thermo Scientific Chemicals

$79.99 - $148.84

Chemical Identifiers

CAS 112883-42-8
Molecular Formula C22H25NO4
Molecular Weight (g/mol) 367.45
MDL Number MFCD00235878
InChI Key RZZXDYZWHUAOEK-GNLPSFAGNA-N
Synonym fmoc-n-me-nle-oh, fmoc-n-methyl-l-norleucine, fmoc-nalpha-methyl-l-norleucine, l-norleucine,n-9h-fluoren-9-ylmethoxy carbonyl-n-methyl, l-norleucine, n-9h-fluoren-9-ylmethoxy carbonyl-n-methyl, 2s-2-9h-fluoren-9-ylmethoxy carbonyl methyl amino hexanoic acid, s-2-9h-fluoren-9-yl methoxy carbonyl methyl amino hexanoic acid, n-alpha-9-fluorenylmethyloxycarbonyl-n-alpha-methyl-l-norleucine, ambotzfaa6460, fmoc-n-me-nle-oh hplc
PubChem CID 7019522
IUPAC Name (2S)-2-[9H-fluoren-9-ylmethoxycarbonyl(methyl)amino]hexanoic acid
SMILES CCCC[C@H](N(C)C(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6399203
View Documents
Thermo Scientific Chemicals
H6399203
1 g
Each for $79.99
 
AAH6399206
View Documents
Thermo Scientific Chemicals
H6399206
5 g
Each for $148.84
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

112883-42-8
367.45
RZZXDYZWHUAOEK-GNLPSFAGNA-N
7019522
CCCC[C@H](N(C)C(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O
C22H25NO4
MFCD00235878
fmoc-n-me-nle-oh, fmoc-n-methyl-l-norleucine, fmoc-nalpha-methyl-l-norleucine, l-norleucine,n-9h-fluoren-9-ylmethoxy carbonyl-n-methyl, l-norleucine, n-9h-fluoren-9-ylmethoxy carbonyl-n-methyl, 2s-2-9h-fluoren-9-ylmethoxy carbonyl methyl amino hexanoic acid, s-2-9h-fluoren-9-yl methoxy carbonyl methyl amino hexanoic acid, n-alpha-9-fluorenylmethyloxycarbonyl-n-alpha-methyl-l-norleucine, ambotzfaa6460, fmoc-n-me-nle-oh hplc
(2S)-2-[9H-fluoren-9-ylmethoxycarbonyl(methyl)amino]hexanoic acid

Specifications

112883-42-8
MFCD00235878
RZZXDYZWHUAOEK-GNLPSFAGNA-N
(2S)-2-[9H-fluoren-9-ylmethoxycarbonyl(methyl)amino]hexanoic acid
367.45
367.44
Solid
C22H25NO4
fmoc-n-me-nle-oh, fmoc-n-methyl-l-norleucine, fmoc-nalpha-methyl-l-norleucine, l-norleucine,n-9h-fluoren-9-ylmethoxy carbonyl-n-methyl, l-norleucine, n-9h-fluoren-9-ylmethoxy carbonyl-n-methyl, 2s-2-9h-fluoren-9-ylmethoxy carbonyl methyl amino hexanoic acid, s-2-9h-fluoren-9-yl methoxy carbonyl methyl amino hexanoic acid, n-alpha-9-fluorenylmethyloxycarbonyl-n-alpha-methyl-l-norleucine, ambotzfaa6460, fmoc-n-me-nle-oh hplc
CCCC[C@H](N(C)C(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O
1 g
7019522
96%
N-Fmoc-N-methyl-L-norleucine
Videos
Safety and Handling

Safety and Handling

TSCA : No

storageNote1 : 2° to 8°C

Recommended Storage : Keep cold

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only