missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N-Fmoc-S-trityl-L-cysteine pentafluorophenyl ester, 98%, Thermo Scientific Chemicals

$110.25 - $382.16

Chemical Identifiers

CAS 115520-21-3
Molecular Formula C43H30F5NO4S
Molecular Weight (g/mol) 751.768
MDL Number MFCD00065639
InChI Key FDAUCYDVXIPBDR-UMSFTDKQSA-N
Synonym fmoc-cys trt-opfp, pentafluorophenyl n-9h-fluoren-9-ylmethoxy carbonyl-s-trityl-l-cysteinate, fmoc-s-trityl-l-cysteine pentafluorophenyl ester, fmoc-s-trityl-l-cysteine pentafluorophenyl ester, fmoc-cys trt-opfp hplc, n, a-fmoc-s-trityl-l-cysteine pentafluorophenyl ester, a-fmoc-s-trityl-l-cysteine pentafluorophenyl ester, n alpha-fmoc-s-trityl-l-cysteine penta-fluorophenyl ester, n alpha-fmoc-s-trityl-l-cysteine penta-fluorophenyl ester, n-alpha-9-fluorenylmethyloxycarbonyl-s-trityl-l-cysteine pentafluorphenyl ester
PubChem CID 11050947
IUPAC Name (2,3,4,5,6-pentafluorophenyl) (2R)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-tritylsulfanylpropanoate
SMILES C1=CC=C(C=C1)C(C2=CC=CC=C2)(C3=CC=CC=C3)SCC(C(=O)OC4=C(C(=C(C(=C4F)F)F)F)F)NC(=O)OCC5C6=CC=CC=C6C7=CC=CC=C57
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6300303
View Documents
Thermo Scientific Chemicals
H6300303
1 g
Each for $110.25
 
AAH6300306
View Documents
Thermo Scientific Chemicals
H6300306
5 g
Each for $382.16
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

115520-21-3
751.768
FDAUCYDVXIPBDR-UMSFTDKQSA-N
11050947
C1=CC=C(C=C1)C(C2=CC=CC=C2)(C3=CC=CC=C3)SCC(C(=O)OC4=C(C(=C(C(=C4F)F)F)F)F)NC(=O)OCC5C6=CC=CC=C6C7=CC=CC=C57
C43H30F5NO4S
MFCD00065639
fmoc-cys trt-opfp, pentafluorophenyl n-9h-fluoren-9-ylmethoxy carbonyl-s-trityl-l-cysteinate, fmoc-s-trityl-l-cysteine pentafluorophenyl ester, fmoc-s-trityl-l-cysteine pentafluorophenyl ester, fmoc-cys trt-opfp hplc, n, a-fmoc-s-trityl-l-cysteine pentafluorophenyl ester, a-fmoc-s-trityl-l-cysteine pentafluorophenyl ester, n alpha-fmoc-s-trityl-l-cysteine penta-fluorophenyl ester, n alpha-fmoc-s-trityl-l-cysteine penta-fluorophenyl ester, n-alpha-9-fluorenylmethyloxycarbonyl-s-trityl-l-cysteine pentafluorphenyl ester
(2,3,4,5,6-pentafluorophenyl) (2R)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-tritylsulfanylpropanoate

Specifications

115520-21-3
MFCD00065639
fmoc-cys trt-opfp, pentafluorophenyl n-9h-fluoren-9-ylmethoxy carbonyl-s-trityl-l-cysteinate, fmoc-s-trityl-l-cysteine pentafluorophenyl ester, fmoc-s-trityl-l-cysteine pentafluorophenyl ester, fmoc-cys trt-opfp hplc, fmoc-cys trt-opfp hplc, a-fmoc-s-trityl-l-cysteine pentafluorophenyl ester, nalpha-fmoc-s-trityl-l-cysteine pentafluorophenyl ester, n alpha-fmoc-s-trityl-l-cysteine penta-fluorophenyl ester, n-alpha-9-fluorenylmethyloxycarbonyl-s-trityl-l-cysteine pentafluorphenyl ester, n-alpha-9-fluorenylmethyloxycarbonyl-s-trityl-l-cysteine pentafluorphenyl ester
C1=CC=C(C=C1)C(C2=CC=CC=C2)(C3=CC=CC=C3)SCC(C(=O)OC4=C(C(=C(C(=C4F)F)F)F)F)NC(=O)OCC5C6=CC=CC=C6C7=CC=CC=C57
1 g
11050947
98%
N-Fmoc-S-trityl-L-cysteine pentafluorophenyl ester
C43H30F5NO4S
4225739
FDAUCYDVXIPBDR-UMSFTDKQSA-N
(2,3,4,5,6-pentafluorophenyl) (2R)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-tritylsulfanylpropanoate
751.768
751.76
Solid
Videos
Safety and Handling

Safety and Handling

TSCA : No

storageNote1 : 2° to 8°C

Recommended Storage : Keep cold

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only