Learn More
N-Formyl-D-leucine, tech. 90%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H3446503
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Spécifications
| N-Formyl-D-leucine | |
| 132°C to 136°C | |
| MFCD08275788 | |
| n-alpha-formyl-d-leucine, 2r-2-formamido-4-methylpentanoic acid, r-2-formamido-4-methylpentanoic acid, formyl-d-leucin, n-formyl-d-leucine, d-leucine, n-formyl, r-2-formylamino-4-methylpentanoic acid | |
| CC(C)C[C@@H](NC=O)C(O)=O | |
| 159.19 | |
| 159.18 | |
| Technical |
| 44978-39-4 | |
| C7H13NO3 | |
| 1 g | |
| HFBHOAHFRNLZGN-MDOHGIEYNA-N | |
| (2R)-2-formamido-4-methylpentanoic acid | |
| 13025456 | |
| 90% |
Identifiants chimiques
| 44978-39-4 | |
| 159.19 | |
| HFBHOAHFRNLZGN-MDOHGIEYNA-N | |
| 13025456 | |
| CC(C)C[C@@H](NC=O)C(O)=O |
| C7H13NO3 | |
| MFCD08275788 | |
| n-alpha-formyl-d-leucine, 2r-2-formamido-4-methylpentanoic acid, r-2-formamido-4-methylpentanoic acid, formyl-d-leucin, n-formyl-d-leucine, d-leucine, n-formyl, r-2-formylamino-4-methylpentanoic acid | |
| (2R)-2-formamido-4-methylpentanoic acid |
Sécurité et manipulation
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only