missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N-Formyl-DL-tryptophan, 98%, Thermo Scientific™

$31.10 - $81.19

Chemical Identifiers

CAS 16108-03-5
Molecular Formula C12H12N2O3
Molecular Weight (g/mol) 232.24
MDL Number MFCD00055983
InChI Key RNEMLJPSSOJRHX-UHFFFAOYSA-N
Synonym n-formyl-dl-tryptophan, n-formyltryptophan, formyl-dl-trp-oh, nalpha-formyl-dl-tryptophan, 2-formamido-3-1h-indol-3-yl propanoic acid, 3-1h-indol-3-yl-2-formamidopropanoic acid, n-formyltryptophan #, tryptophan, n-formyl, for-dl-trp-oh, acmc-1clw6
PubChem CID 582651
IUPAC Name 2-formamido-3-(1H-indol-3-yl)propanoic acid
SMILES OC(=O)C(CC1=CNC2=CC=CC=C12)NC=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH63446MD
View Documents
Thermo Scientific Chemicals
H63446MD
250 mg
Each for $31.10
 
AAH6344603
View Documents
Thermo Scientific Chemicals
H6344603
1 g
Each for $81.19
 
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

16108-03-5
232.24
RNEMLJPSSOJRHX-UHFFFAOYSA-N
582651
OC(=O)C(CC1=CNC2=CC=CC=C12)NC=O
C12H12N2O3
MFCD00055983
n-formyl-dl-tryptophan, n-formyltryptophan, formyl-dl-trp-oh, nalpha-formyl-dl-tryptophan, 2-formamido-3-1h-indol-3-yl propanoic acid, 3-1h-indol-3-yl-2-formamidopropanoic acid, n-formyltryptophan #, tryptophan, n-formyl, for-dl-trp-oh, acmc-1clw6
2-formamido-3-(1H-indol-3-yl)propanoic acid

Specifications

16108-03-5
MFCD00055983
RNEMLJPSSOJRHX-UHFFFAOYSA-N
2-formamido-3-(1H-indol-3-yl)propanoic acid
232.24
232.24
N-Formyl-DL-tryptophan
C12H12N2O3
n-formyl-dl-tryptophan, n-formyltryptophan, formyl-dl-trp-oh, nalpha-formyl-dl-tryptophan, 2-formamido-3-1h-indol-3-yl propanoic acid, 3-1h-indol-3-yl-2-formamidopropanoic acid, n-formyltryptophan #, tryptophan, n-formyl, for-dl-trp-oh, acmc-1clw6
OC(=O)C(CC1=CNC2=CC=CC=C12)NC=O
250 mg
582651
98%
Videos
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'