missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N-Formylpiperidine, 99%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 119732500

 View more versions of this product

Catalog No. AC119732500


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

N-Formylpiperidine
2591-86-8
100.0
Colorless to Yellow
222°C
Authentic
Glass bottle
1.4830 to 1.485
250 mL
11,244
n-formylpiperidine, 1-formylpiperidine, 1-piperidinecarboxaldehyde, formylpiperidine, n-formylpiperidin, piperidinoformamide, piperidine-n-carbaldehyde, piperidine, 1-formyl, 1-piperidinecarbaldehyde, unii-ziq29h6czg
FEWLNYSYJNLUOO-UHFFFAOYSA-N
piperidine-1-carbaldehyde
17429
113.16
Liquid
99%
98.5
-31°C
1.0190g/mL
91°C
99%
C6H11NO
MFCD00006483
20, 45
1.019
Solubility in water: freely soluble. Other solubilities: soluble in ethanol,ether,hexane
C1CCN(CC1)C=O
113.16
CHEBI:42546
99%

Chemical Identifiers

2591-86-8
113.16
FEWLNYSYJNLUOO-UHFFFAOYSA-N
17429
piperidine-1-carbaldehyde
C6H11NO
MFCD00006483
n-formylpiperidine, 1-formylpiperidine, 1-piperidinecarboxaldehyde, formylpiperidine, n-formylpiperidin, piperidinoformamide, piperidine-n-carbaldehyde, piperidine, 1-formyl, 1-piperidinecarbaldehyde, unii-ziq29h6czg
CHEBI:42546
C1CCN(CC1)C=O
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
Toxic in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Do not eat,drink or smoke when using this product.

GHS Signal Word: Danger

EINECSNumber : 219-986-0

RTECSNumber : TN0380000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only