Learn More
n-Heptane, 99.5%, for analysis, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 389170025
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For analysisSpecifications
n-Heptane | |
142-82-5 | |
100.0 | |
0.6830g/mL | |
2.5 L | |
Glass Bottle | |
1.3850 to 1.3890 | |
99.5% | |
C7H16 | |
MFCD00009544 | |
0.02% max. (K.F.) | |
Solubility in water: practically insoluble. Other solubilities: soluble in alcohol, ether and chloroform | |
CCCCCCC | |
100.21 | |
CHEBI:43098 | |
99.5% |
-91.0°C | |
99.4 | |
0.0001 meq/g max. | |
98.0°C | |
−4°C | |
Liquid | |
0.001% max. | |
0.683 | |
CH3(CH2)5CH3 | |
0.4 mPa.s (20°C) | |
n-heptane, dipropylmethane, heptan, heptyl hydride, dipropyl methane, gettysolve-c, skellysolve c, heptanen, eptani, heptan polish | |
IMNFDUFMRHMDMM-UHFFFAOYSA-N | |
heptane | |
8900 | |
100.2 | |
Analytical |
Chemical Identifiers
142-82-5 | |
100.21 | |
IMNFDUFMRHMDMM-UHFFFAOYSA-N | |
8900 | |
heptane |
C7H16 | |
MFCD00009544 | |
n-heptane, dipropylmethane, heptan, heptyl hydride, dipropyl methane, gettysolve-c, skellysolve c, heptanen, eptani, heptan polish | |
CHEBI:43098 | |
CCCCCCC |
Safety and Handling
GHS H Statement
Causes skin irritation. May cause drowsiness or dizziness. May be fatal if swallowed and enters airways. Highly flammable liquid and vapour. Very toxic to aquatic life with long lasting effects.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray. IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician. Do NOT induce vomiting. Avoid release to the environment. IF ON SKIN: Wash with plenty of soap and water. Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Danger
RUO – Research Use Only