Learn More
n-Heptane, 99+%, Extra Dry over Molecular Sieve, AcroSeal™, Thermo Scientific Chemicals
$84.33 - $355.73
Chemical Identifiers
CAS | 142-82-5 |
---|---|
Molecular Formula | C7H16 |
Molecular Weight (g/mol) | 100.21 |
MDL Number | MFCD00009544 |
InChI Key | IMNFDUFMRHMDMM-UHFFFAOYSA-N |
Synonym | n-heptane, dipropylmethane, heptan, heptyl hydride, dipropyl methane, gettysolve-c, skellysolve c, heptanen, eptani, heptan polish |
PubChem CID | 8900 |
ChEBI | CHEBI:43098 |
IUPAC Name | heptane |
SMILES | CCCCCCC |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC364361000
|
Thermo Scientific Chemicals
364361000 |
100 mL | AcroSeal™ Glass Bottle |
Each for $84.33
|
|
||||
AC364360010
|
Thermo Scientific Chemicals
364360010 |
1 L | AcroSeal™ Glass Bottle |
Each for $154.99
|
|
||||
AC364360025
|
Thermo Scientific Chemicals
364360025 |
2.5 L | AcroSeal™ Glass Bottle |
Each for $355.73
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
142-82-5 | |
100.21 | |
IMNFDUFMRHMDMM-UHFFFAOYSA-N | |
8900 | |
heptane |
C7H16 | |
MFCD00009544 | |
n-heptane, dipropylmethane, heptan, heptyl hydride, dipropyl methane, gettysolve-c, skellysolve c, heptanen, eptani, heptan polish | |
CHEBI:43098 | |
CCCCCCC |
Specifications
-91.0°C | |
Colorless | |
100.0 | |
98.0°C | |
−4°C | |
AcroSeal™ Glass Bottle | |
1.3850 to 1.3890 | |
0.683 | |
CH3(CH2)5CH3 | |
0.4 mPa.s (20°C) | |
01, 154 | |
n-heptane, dipropylmethane, heptan, heptyl hydride, dipropyl methane, gettysolve-c, skellysolve c, heptanen, eptani, heptan polish | |
IMNFDUFMRHMDMM-UHFFFAOYSA-N | |
heptane | |
8900 | |
100.2 | |
Extra Dry over Molecular Sieve |
142-82-5 | |
99.0 | |
0.6830g/mL | |
100 mL | |
Authentic | |
Liquid | |
99+% | |
C7H16 | |
MFCD00009544 | |
0.005% Max. | |
15, 4694 | |
Solubility in water: practically insoluble. Other solubilities: soluble in alcohol, ether and chloroform | |
CCCCCCC | |
100.21 | |
CHEBI:43098 | |
≥99% | |
n-Heptane, Over Molecular Sieves |
Safety and Handling
GHS H Statement
Causes skin irritation. May cause drowsiness or dizziness. May be fatal if swallowed and enters airways. Highly flammable liquid and vapour. Very toxic to aquatic life with long lasting effects.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray. IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician. Do NOT induce vomiting. Avoid release to the environment. IF ON SKIN: Wash with plenty of soap and water. Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Danger
EINECSNumber : 205-563-8
RTECSNumber : MI7700000
TSCA : TSCA
RUO – Research Use Only