missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

n-Heptane, 99+%, extra pure, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 383790250

 View more versions of this product

Catalog No. AC383790250

Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

n-Heptane
142-82-5
0.003 meq/g max.
98.0°C
−4°C
Liquid
20ppm max.
0.683
CH3(CH2)5CH3
0.4 mPa.s (20°C)
15, 4694
Solubility in water: practically insoluble. Other solubilities: soluble in alcohol, ether and chloroform
CCCCCCC
100.21
CHEBI:43098
≥99%
-91.0°C
Colorless to Yellow
0.6830g/mL
25 L
Authentic
1.3850 to 1.3890
99+%
C7H16
MFCD00009544
0.05% max.
n-heptane, dipropylmethane, heptan, heptyl hydride, dipropyl methane, gettysolve-c, skellysolve c, heptanen, eptani, heptan polish
IMNFDUFMRHMDMM-UHFFFAOYSA-N
heptane
8900
100.2
Extra Pure

Chemical Identifiers

142-82-5
100.21
IMNFDUFMRHMDMM-UHFFFAOYSA-N
8900
heptane
C7H16
MFCD00009544
n-heptane, dipropylmethane, heptan, heptyl hydride, dipropyl methane, gettysolve-c, skellysolve c, heptanen, eptani, heptan polish
CHEBI:43098
CCCCCCC
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation. May cause drowsiness or dizziness. May be fatal if swallowed and enters airways. Highly flammable liquid and vapour. Very toxic to aquatic life with long lasting effects.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray. IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician. Do NOT induce vomiting. Avoid release to the environment. IF ON SKIN: Wash with plenty of soap and water. Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

Danger

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only