missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

n-Heptane, 99%, Optima™ for HPLC and GC, Fisher Chemical

High purity and versatile solvent suitable for HPLC, spectrophotometry, and pesticide residue analysis

Supplier:  Fisher Chemical H3601

Catalog No. H3601



Explore available promotions

Description

Description

  • Screened for fluorescing contaminants, and UV-absorbing contaminants in the 197-300nm range, to reduce interferences for smoother baselines
  • Tested by GC-ECD to ensure minimum halogenated residue levels (≤10 ng/L) to demonstrate suitability for pesticide analysis
  • Filtered through a sub-micron filter to remove particulates
  • Intended for research use only
Specifications

Specifications

n-Heptane
142-82-5
Colorless
98°C
-4°C (25°F)
Amber Soda Lime Glass
1.3845 to 1.3865 (at 25°C)
99.0%
CH3(CH2)5CH3
48mbar at 20°C
0.01% max.
IMNFDUFMRHMDMM-UHFFFAOYSA-N
heptane
8900
100.2
-91°C
0.005 AU max. at 270nm, 0.005 AU max. at 300nm, 0.01 AU max. at 250nm, 0.01 AU max. at 260nm, 0.05 AU max. at 230nm, 0.10 AU max. at 220nm, 0.10 AU max. at 225nm, 0.25 AU max. at 210nm, 1.00 AU max. at 197nm
0.683g/cm³
1 L
Pass Test
Liquid
1ppm max.
C7H16
MFCD00009544
0.4 mPa/s at 20°C
n-heptane, dipropylmethane, heptan, heptyl hydride, dipropyl methane, gettysolve-c, skellysolve c, heptanen, eptani, heptan polish
CCCCCCC
100.21
CHEBI:43098
OPTIMA™
Videos
Safety and Handling

Safety and Handling

DANGER!

Emergency Overview
Flammable liquid and vapor. Inhalation may cause central nervous system effects. May cause irritation of respiratory tract. Aspiration hazard if swallowed - can enter lungs and cause damage. Very toxic to aquatic organisms, may cause long-term adverse effects in the aquatic environment. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Obtain medical attention. Move to fresh air.

NFPA

Health:1
Flammability:3
Instability:0

DOTInformation : DOT Class 3, : Flammable Liquid

SDS Documents

Documents

Accessories
Promotions

Promotions