missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

n-Heptane, 99+%, residue analysis, for trace analysis of polyaromatic hydrocarb, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 326770025

 View more versions of this product

Catalog No. AC326770025


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For residue analysis
Specifications

Specifications

n-Heptane
Tested for Trace Analysis of Polyaromatic Hydrocarbons
99.0
0.002% max.
98.0°C
−4°C
1.3850 to 1.3890
99+%
C7H16
MFCD00009544
0.001% max. (Coulometric)
15, 4694
Solubility in water: practically insoluble. Other solubilities: soluble in alcohol, ether and chloroform
CCCCCCC
100.21
CHEBI:43098
≥99%
-91.0°C
142-82-5
100.0
0.6830g/mL
2.5 L
Liquid
0.0003% max.
0.683
CH3(CH2)5CH3
0.4 mPa.s (20°C)
01, 154
n-heptane, dipropylmethane, heptan, heptyl hydride, dipropyl methane, gettysolve-c, skellysolve c, heptanen, eptani, heptan polish
IMNFDUFMRHMDMM-UHFFFAOYSA-N
heptane
8900
100.2
Residue Analysis

Chemical Identifiers

142-82-5
100.21
IMNFDUFMRHMDMM-UHFFFAOYSA-N
8900
heptane
C7H16
MFCD00009544
n-heptane, dipropylmethane, heptan, heptyl hydride, dipropyl methane, gettysolve-c, skellysolve c, heptanen, eptani, heptan polish
CHEBI:43098
CCCCCCC
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation. May cause drowsiness or dizziness. May be fatal if swallowed and enters airways. Highly flammable liquid and vapour. Very toxic to aquatic life with long lasting effects.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray. IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician. Do NOT induce vomiting. Avoid release to the environment. IF ON SKIN: Wash with plenty of soap and water. Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

Danger

EINECSNumber : 205-563-8

RTECSNumber : MI7700000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only