missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

n-Hexane For gas chromatography ECD and FID SupraSolv™, MilliporeSigma™

For gas chromatography ECD and FID SupraSolv™

$201.24 - $426.76

Chemical Identifiers

CAS 110-54-3
Molecular Formula C6H14
Molecular Weight (g/mol) 86.18
MDL Number MFCD02179311
InChI Key VLKZOEOYAKHREP-UHFFFAOYSA-N
Synonym n-hexane, esani, skellysolve b, gettysolve-b, hexyl hydride, hexan, dipropyl, heksan, hexanen, hexanes
PubChem CID 8058
ChEBI CHEBI:29021
IUPAC Name hexane
SMILES CCCCCC
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
M1043711000
View Documents
Technidata
1043711000
1 L Glass Bottle
Each for $201.24
 
M1043712500
View Documents
Technidata
1043712500
2.5 L Glass Bottle
Each for $426.76
 
Description

Description

Specifications

Chemical Identifiers

110-54-3
86.18
VLKZOEOYAKHREP-UHFFFAOYSA-N
8058
hexane
C6H14
MFCD02179311
n-hexane, esani, skellysolve b, gettysolve-b, hexyl hydride, hexan, dipropyl, heksan, hexanen, hexanes
CHEBI:29021
CCCCCC

Specifications

-94.3°C
Colorless
69°C
−22°C (−7.6°F)
Benzine-like
Liquid
C6H14
160 hPa (20°C)
n-hexane, esani, skellysolve b, gettysolve-b, hexyl hydride, hexan, dipropyl, heksan, hexanen, hexanes
VLKZOEOYAKHREP-UHFFFAOYSA-N
hexane
8058
86.18
110-54-3
0.66g/cm3 at 20°C
1 L
240° (464°F)
Glass Bottle
1.37
MFCD02179311
1208
Soluble in water: 0.0095g/L at 20°C
CCCCCC
86.18
CHEBI:29021
Hexane
Videos
Safety and Handling

Safety and Handling

H304: May be fatal if swallowed and enters airways.

H315: Causes skin irritation.

H225: Highly flammable liquid and vapour.

Recommended Storage : 5°C to 30°C (41°F to 86°F); Keep container tightly closed in a dry and well-ventilated place. Keep away from heat and sources of ignition.

SDS
missing translation for 'documents'

missing translation for 'documents'