Learn More
n-Hexylphosphonic acid, Thermo Scientific Chemicals
$202.71 - $732.30
Chemical Identifiers
CAS | 4721-24-8 |
---|---|
Molecular Formula | C6H15O3P |
Molecular Weight (g/mol) | 166.157 |
MDL Number | MFCD00013961 |
InChI Key | GJWAEWLHSDGBGG-UHFFFAOYSA-N |
Synonym | n-hexylphosphonic acid, phosphonic acid, p-hexyl, hexylphosphonicacid, p-hexyl-phosphonic acid, hexylphosphonic acid |
PubChem CID | 312552 |
IUPAC Name | hexylphosphonic acid |
SMILES | CCCCCCP(=O)(O)O |
Description
n-Hexylphosphonic acid is used in the manufacturing of nanoparticles such as quantum dots, nano-metals and nano-ceramics. It is also used for coating of many materials, including nanoparticles by condensed hydrophobic monolayers.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
4721-24-8 | |
166.157 | |
GJWAEWLHSDGBGG-UHFFFAOYSA-N | |
312552 | |
CCCCCCP(=O)(O)O |
C6H15O3P | |
MFCD00013961 | |
n-hexylphosphonic acid, phosphonic acid, p-hexyl, hexylphosphonicacid, p-hexyl-phosphonic acid, hexylphosphonic acid | |
hexylphosphonic acid |
Specifications
4721-24-8 | |
White | |
MFCD00013961 | |
n-hexylphosphonic acid, phosphonic acid, p-hexyl, hexylphosphonicacid, p-hexyl-phosphonic acid, hexylphosphonic acid | |
GJWAEWLHSDGBGG-UHFFFAOYSA-N | |
hexylphosphonic acid | |
312552 | |
Powder |
108°C to 110°C | |
C6H15O3P | |
2 g | |
Insoluble in water. | |
CCCCCCP(=O)(O)O | |
166.157 | |
166.16 | |
n-Hexylphosphonic acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only