missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
N-Hydroxy-4-nitrophthalimide 98.0+%, TCI America™
Supplier: TCI America H10365G
Specifications
N-Hydroxy-4-nitrophthalimide | |
169°C | |
C8H4N2O5 | |
5 g | |
MKACMVMZUIQKNY-UHFFFAOYSA-N | |
2-hydroxy-5-nitro-2,3-dihydro-1H-isoindole-1,3-dione | |
11458432 | |
≥98.0% (HPLC) |
105969-98-0 | |
White-Yellow | |
MFCD01115771 | |
4-Nitrophthaloxime | |
ON1C(=O)C2=C(C=C(C=C2)[N+]([O-])=O)C1=O | |
208.13 | |
208.13 | |
Crystalline Powder |
Chemical Identifiers
105969-98-0 | |
208.13 | |
MKACMVMZUIQKNY-UHFFFAOYSA-N | |
11458432 | |
ON1C(=O)C2=C(C=C(C=C2)[N+]([O-])=O)C1=O |
C8H4N2O5 | |
MFCD01115771 | |
4-Nitrophthaloxime | |
2-hydroxy-5-nitro-2,3-dihydro-1H-isoindole-1,3-dione |
Safety and Handling
TSCA : No