missing translation for 'onlineSavingsMsg'
Learn More

N-(Hydroxymethyl)acetamide, 98%

CAS: 625-51-4 | C3H7NO2 | 89.09 g/mol

$112.10 - $201.25

Chemical Identifiers

CAS 625-51-4
Molecular Formula C3H7NO2
Molecular Weight (g/mol) 89.09
MDL Number MFCD00014417
InChI Key HWJHZLJIIWOTGZ-UHFFFAOYSA-N
Synonym n-hydroxymethyl acetamide, acetamidomethanol, formicin, acetamide, n-hydroxymethyl, unii-rcg03a51mo, n-hydroxymethyl-acetamide, rcg03a51mo, acetamido methanol, n-acetylmethanolamine, acetylamino methanol
PubChem CID 69365
IUPAC Name N-(hydroxymethyl)acetamide
SMILES CC(=O)NCO
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1382009
View Documents
Thermo Scientific Chemicals
A1382009
10 g
Each for $112.10
Only null left
 
AAA1382014
View Documents
Thermo Scientific Chemicals
A1382014
25 g
Each for $201.25
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

625-51-4
89.09
HWJHZLJIIWOTGZ-UHFFFAOYSA-N
69365
CC(=O)NCO
C3H7NO2
MFCD00014417
n-hydroxymethyl acetamide, acetamidomethanol, formicin, acetamide, n-hydroxymethyl, unii-rcg03a51mo, n-hydroxymethyl-acetamide, rcg03a51mo, acetamido methanol, n-acetylmethanolamine, acetylamino methanol
N-(hydroxymethyl)acetamide

Specifications

625-51-4
1.14
MFCD00014417
506226
14,4242
HWJHZLJIIWOTGZ-UHFFFAOYSA-N
N-(hydroxymethyl)acetamide
69365
98%
47°C to 52°C
C3H7NO2
10 g
Hygroscopic
n-hydroxymethyl acetamide, acetamidomethanol, formicin, acetamide, n-hydroxymethyl, unii-rcg03a51mo, n-hydroxymethyl-acetamide, rcg03a51mo, acetamido methanol, n-acetylmethanolamine, acetylamino methanol
CC(=O)NCO
89.09
89.09
N-(Hydroxymethyl)acetamide
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 210-897-2

RTECSNumber : AC3610000

TSCA : No

Recommended Storage : Keep cold

SDS
Documents

Documents

RUO – Research Use Only