missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N-(Hydroxymethyl)acrylamide 98.0+%, TCI America™

Supplier:  TCI America M057425G

 View more versions of this product

Catalog No. M057425G


Explore available promotions

Description

Description

Specifications

Specifications

N-(Hydroxymethyl)acrylamide
76°C
C4H7NO2
25 g
n-methylolacrylamide, n-hydroxymethyl acrylamide, n-methanolacrylamide, monomethylolacrylamide, methylolacrylamide, n-methylol acrylamide, 2-propenamide, n-hydroxymethyl, acrylamide, n-hydroxymethyl, uramine t 80, n-hydroxymethyl-2-propenamide
C=CC(=O)NCO
101.105
CHEBI:82492
≥98.0% (T)
924-42-5
White
MFCD00004597
3531
CNCOEDDPFOAUMB-UHFFFAOYSA-N
N-(hydroxymethyl)prop-2-enamide
13543
101.11
Crystalline Powder

Chemical Identifiers

924-42-5
101.105
CNCOEDDPFOAUMB-UHFFFAOYSA-N
13543
N-(hydroxymethyl)prop-2-enamide
C4H7NO2
MFCD00004597
n-methylolacrylamide, n-hydroxymethyl acrylamide, n-methanolacrylamide, monomethylolacrylamide, methylolacrylamide, n-methylol acrylamide, 2-propenamide, n-hydroxymethyl, acrylamide, n-hydroxymethyl, uramine t 80, n-hydroxymethyl-2-propenamide
CHEBI:82492
C=CC(=O)NCO
Safety and Handling

Safety and Handling

EINECSNumber : (2)-1022

RTECSNumber : AS3600000

TSCA : Yes

Recommended Storage : Refrigerator

SDS
Documents

Documents

Promotions

Promotions