Learn More
N-Isopropyl-4-nitroaniline, 97+%, Thermo Scientific™
$143.90 - $143.90
Chemical Identifiers
CAS | 25186-43-0 |
---|---|
Molecular Formula | C9H12N2O2 |
Molecular Weight (g/mol) | 180.207 |
MDL Number | MFCD00128997 |
InChI Key | VTSUWHFLMJLYKN-UHFFFAOYSA-N |
Synonym | n-isopropyl-4-nitroaniline, benzenamine, n-1-methylethyl-4-nitro, 4-isopropylamino-nitrobenzene, p-nitrophenyl-isopropylamine, 4-nitro-n-propan-2-yl aniline, n-1-methylethyl-4-nitroaniline, benzenamine,n-1-methylethyl-4-nitro |
PubChem CID | 32833 |
IUPAC Name | 4-nitro-N-propan-2-ylaniline |
SMILES | CC(C)NC1=CC=C(C=C1)[N+](=O)[O-] |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAH5237503
|
Thermo Scientific Chemicals
H5237503 |
1 g |
Each for $143.90
|
|
|||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
25186-43-0 | |
180.207 | |
VTSUWHFLMJLYKN-UHFFFAOYSA-N | |
32833 | |
CC(C)NC1=CC=C(C=C1)[N+](=O)[O-] |
C9H12N2O2 | |
MFCD00128997 | |
n-isopropyl-4-nitroaniline, benzenamine, n-1-methylethyl-4-nitro, 4-isopropylamino-nitrobenzene, p-nitrophenyl-isopropylamine, 4-nitro-n-propan-2-yl aniline, n-1-methylethyl-4-nitroaniline, benzenamine,n-1-methylethyl-4-nitro | |
4-nitro-N-propan-2-ylaniline |
Specifications
25186-43-0 | |
MFCD00128997 | |
UN2811 | |
VTSUWHFLMJLYKN-UHFFFAOYSA-N | |
4-nitro-N-propan-2-ylaniline | |
32833 | |
≥97% |
C9H12N2O2 | |
1 g | |
n-isopropyl-4-nitroaniline, benzenamine, n-1-methylethyl-4-nitro, 4-isopropylamino-nitrobenzene, p-nitrophenyl-isopropylamine, 4-nitro-n-propan-2-yl aniline, n-1-methylethyl-4-nitroaniline, benzenamine,n-1-methylethyl-4-nitro | |
CC(C)NC1=CC=C(C=C1)[N+](=O)[O-] | |
180.207 | |
180.2 | |
N-Isopropyl-4-nitroaniline |
Safety and Handling
GHS H Statement
H331-H302-H312
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c
H302+H312+H332
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber : 246-721-6
TSCA : Yes
Recommended Storage : Ambient temperatures