missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
N'-Isopropylidene-2-nitrobenzenesulfonohydrazide 98.0+%, TCI America™
$110.60 - $345.62
Chemical Identifiers
CAS | 6655-27-2 |
---|---|
Molecular Formula | C9H11N3O4S |
Molecular Weight (g/mol) | 257.264 |
MDL Number | MFCD09800525 |
InChI Key | SBNYNTYNEJTMQO-UHFFFAOYSA-N |
Synonym | IPNBSH |
PubChem CID | 11974266 |
SMILES | CC(=NNS(=O)(=O)C1=CC=CC=C1[N+](=O)[O-])C |
Chemical Identifiers
6655-27-2 | |
257.264 | |
SBNYNTYNEJTMQO-UHFFFAOYSA-N | |
11974266 |
C9H11N3O4S | |
MFCD09800525 | |
IPNBSH | |
CC(=NNS(=O)(=O)C1=CC=CC=C1[N+](=O)[O-])C |
Specifications
6655-27-2 | |
C9H11N3O4S | |
1 g | |
SBNYNTYNEJTMQO-UHFFFAOYSA-N | |
257.264 | |
257.26 | |
Crystalline Powder |
136°C | |
MFCD09800525 | |
IPNBSH | |
CC(=NNS(=O)(=O)C1=CC=CC=C1[N+](=O)[O-])C | |
11974266 | |
≥98.0% (HPLC,T) | |
N′-Isopropylidene-2-nitrobenzenesulfonohydrazide |
Safety and Handling
TSCA : No
Recommended Storage : Freezer