Learn More
N-Isopropylpropylamine, 96%, Thermo Scientific Chemicals
$560.22 - $560.22
Chemical Identifiers
CAS | 21968-17-2 |
---|---|
Molecular Formula | C6H15N |
Molecular Weight (g/mol) | 101.193 |
MDL Number | MFCD00027095 |
InChI Key | VLSTXUUYLIALPB-UHFFFAOYSA-N |
Synonym | n-isopropylpropylamine, propan-2-yl propyl amine, n-propyl-n-isopropylamine, unii-k1p2fy26ro, k1p2fy26ro, 1-propanamine, n-1-methylethyl, propyl-isopropylamine, acmc-20ak2r, n-isopropyl-n-propylamine |
PubChem CID | 89119 |
IUPAC Name | N-propan-2-ylpropan-1-amine |
SMILES | CCCNC(C)C |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAH5025603
|
Thermo Scientific Chemicals
H5025603 |
1 g |
Each for $560.22
|
|
|||||
Description
N-Isopropylpropylamine is used as an active pharmaceutical ingredients.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsN-Isopropylpropylamine is used as an active pharmaceutical ingredients.
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Chemical Identifiers
21968-17-2 | |
101.193 | |
VLSTXUUYLIALPB-UHFFFAOYSA-N | |
89119 | |
CCCNC(C)C |
C6H15N | |
MFCD00027095 | |
n-isopropylpropylamine, propan-2-yl propyl amine, n-propyl-n-isopropylamine, unii-k1p2fy26ro, k1p2fy26ro, 1-propanamine, n-1-methylethyl, propyl-isopropylamine, acmc-20ak2r, n-isopropyl-n-propylamine | |
N-propan-2-ylpropan-1-amine |
Specifications
21968-17-2 | |
CH3CH2CH2NHCH(CH3)2 | |
1 g | |
n-isopropylpropylamine, propan-2-yl propyl amine, n-propyl-n-isopropylamine, unii-k1p2fy26ro, k1p2fy26ro, 1-propanamine, n-1-methylethyl, propyl-isopropylamine, acmc-20ak2r, n-isopropyl-n-propylamine | |
CCCNC(C)C | |
101.193 | |
101.19 | |
N-Isopropylpropylamine |
C6H15N | |
MFCD00027095 | |
UN2733 | |
VLSTXUUYLIALPB-UHFFFAOYSA-N | |
N-propan-2-ylpropan-1-amine | |
89119 | |
96% |
Safety and Handling
GHS H Statement
H224-H314-H318
Extremely flammable liquid and vapor.
Causes severe skin burns and eye damage.
Causes serious eye damage.
P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
H225-H314-H335
DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: AMINES, FLAMMABLE, CORROSIVE, N.O.S.
EINECSNumber : 244-688-2
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only