missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N-Methoxy-N-methylacetamide, 98%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 369310250

Catalog No. AC369310250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

N-Methoxy-N-methylacetamide
78191-00-1
100.0
152°C
Authentic
Glass bottle
1.4240 to 1.426
MFCD00060098
0.97
Solubility in water: soluble
CON(C)C(C)=O
103.12
103.12
98%
97.5
0.9700g/mL
49°C
97.5% min. (GC)
C4H9NO2
CH3CON(OCH3)CH3
25g
n-methyl-n-methoxyacetamide, acetamide, n-methoxy-n-methyl, n-methoxyl-n-methylacetamide, n-methoxy-n-methyl-acetamide, n-methoxy-n-methylacetamid, pubchem12775, acmc-209pdo, n-methoxy-n-methylacetamde, n-methoxy n-methylacetamide, n-methoxy-n-methyl acetamide
OYVXVLSZQHSNDK-UHFFFAOYSA-N
N-methoxy-N-methylacetamide
537505
98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Flammable liquid and vapor.

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only