missing translation for 'onlineSavingsMsg'
Learn More

N-Methyl-1,2-phenylenediamine, 97%, Thermo Scientific™

Chemical Identifiers

CAS 4760-34-3
Molecular Formula C7H10N2
Molecular Weight (g/mol) 122.17
MDL Number MFCD00192314
InChI Key RPKCLSMBVQLWIN-UHFFFAOYSA-N
Synonym n1-methylbenzene-1,2-diamine, n-methyl-o-phenylenediamine, n-methylbenzene-1,2-diamine, 1,2-benzenediamine, n-methyl, n-methyl-1,2-phenylenediamine, n-methyl-1,2-benzenediamine, 2-amino-n-methylaniline, n-methyl-2-phenylenediamine, 2-methylamino aniline, unii-roi30fpr50
PubChem CID 78498
IUPAC Name 2-N-methylbenzene-1,2-diamine
SMILES CNC1=CC=CC=C1N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC434230050
SDS View Documents
Thermo Scientific Chemicals
434230050
5mL Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC434230250
SDS View Documents
Thermo Scientific Chemicals
434230250
25mL Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

4760-34-3
122.17
RPKCLSMBVQLWIN-UHFFFAOYSA-N
78498
CNC1=CC=CC=C1N
C7H10N2
MFCD00192314
n1-methylbenzene-1,2-diamine, n-methyl-o-phenylenediamine, n-methylbenzene-1,2-diamine, 1,2-benzenediamine, n-methyl, n-methyl-1,2-phenylenediamine, n-methyl-1,2-benzenediamine, 2-amino-n-methylaniline, n-methyl-2-phenylenediamine, 2-methylamino aniline, unii-roi30fpr50
2-N-methylbenzene-1,2-diamine

Specifications

4760-34-3
100.0
123°C to 124°C (10.0mmHg)
Authentic
Glass bottle
MFCD00192314
1.075
RPKCLSMBVQLWIN-UHFFFAOYSA-N
2-N-methylbenzene-1,2-diamine
78498
97%
96.0
1.0750g/mL
113°C
96% min. (GC)
C7H10N2
5mL
n1-methylbenzene-1,2-diamine, n-methyl-o-phenylenediamine, n-methylbenzene-1,2-diamine, 1,2-benzenediamine, n-methyl, n-methyl-1,2-phenylenediamine, n-methyl-1,2-benzenediamine, 2-amino-n-methylaniline, n-methyl-2-phenylenediamine, 2-methylamino aniline, unii-roi30fpr50
CNC1=CC=CC=C1N
122.17
122.17
N-Methyl-1, 2-phenylenediamine
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if inhaled.
Harmful in contact with skin.
Harmful if swallowed.
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 225-297-6

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only