missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N-Methyl-1,3-propanediamine, 99%, Thermo Scientific Chemicals

$354.27 - $354.27

Chemical Identifiers

CAS 6291-84-5
Molecular Formula C4H12N2
Molecular Weight (g/mol) 88.154
MDL Number MFCD00008209
InChI Key QHJABUZHRJTCAR-UHFFFAOYSA-N
Synonym n-methyl-1,3-propanediamine, n-methyl-1,3-diaminopropane, 3-methylamino propylamine, 1,3-propanediamine, n-methyl, n-methylpropane-1,3-diamine, 3-aminopropyl methylamine, n-methyltrimethylenediamine, 3-aminopropylmethylamine, n-methyl-1,3-propylenediamine, 1-amino-3-methylamino propane
PubChem CID 80511
IUPAC Name N'-methylpropane-1,3-diamine
SMILES CNCCCN
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAB2331936
View Documents
Thermo Scientific Chemicals
B2331936
500 g
Each for $354.27
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

6291-84-5
88.154
QHJABUZHRJTCAR-UHFFFAOYSA-N
80511
CNCCCN
C4H12N2
MFCD00008209
n-methyl-1,3-propanediamine, n-methyl-1,3-diaminopropane, 3-methylamino propylamine, 1,3-propanediamine, n-methyl, n-methylpropane-1,3-diamine, 3-aminopropyl methylamine, n-methyltrimethylenediamine, 3-aminopropylmethylamine, n-methyl-1,3-propylenediamine, 1-amino-3-methylamino propane
N'-methylpropane-1,3-diamine

Specifications

6291-84-5
0.844
35°C (96°F)
1.4475
500 g
878143
n-methyl-1,3-propanediamine, n-methyl-1,3-diaminopropane, 3-methylamino propylamine, 1,3-propanediamine, n-methyl, n-methylpropane-1,3-diamine, 3-aminopropyl methylamine, n-methyltrimethylenediamine, 3-aminopropylmethylamine, n-methyl-1,3-propylenediamine, 1-amino-3-methylamino propane
CNCCCN
88.154
88.15
N-Methyl-1,3-propanediamine
-72°C
139°C to 141°C
C4H12N2
MFCD00008209
UN2734
Air Sensitive
QHJABUZHRJTCAR-UHFFFAOYSA-N
N'-methylpropane-1,3-diamine
80511
99%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H314-H318-H226
Toxic if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapour.

P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P284-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c

H226-H302-H314-H330-H335

DOTInformation : Hazard Class: 8; Packaging Group: I

EINECSNumber : 228-544-6

RTECSNumber : TX8242500

TSCA : Yes

Recommended Storage : Ambient temperatures; Store under Argon

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only