missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N-Methyl-1-propylamine, 97%, Thermo Scientific Chemicals

$61.73 - $613.59

Chemical Identifiers

CAS 627-35-0
Molecular Formula C4H11N
Molecular Weight (g/mol) 73.139
MDL Number MFCD00009361
InChI Key GVWISOJSERXQBM-UHFFFAOYSA-N
Synonym n-methylpropylamine, n-methyl-n-propylamine, methyl propyl amine, 1-propanamine, n-methyl, methyl-n-propylamine, methylpropylamine, unii-vw1h32h1qm, n-propylmethylamine, n-methyl-1-propylamine, vw1h32h1qm
PubChem CID 12315
IUPAC Name N-methylpropan-1-amine
SMILES CCCNC
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1690303
View Documents
Thermo Scientific Chemicals
A1690303
1 g
Each for $61.73
 
AAA1690306
View Documents
Thermo Scientific Chemicals
A1690306
5 g
Each for $131.32
 
AAA1690314
View Documents
Thermo Scientific Chemicals
A1690314
25 g
Each for $613.59
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

627-35-0
73.139
GVWISOJSERXQBM-UHFFFAOYSA-N
12315
CCCNC
C4H11N
MFCD00009361
n-methylpropylamine, n-methyl-n-propylamine, methyl propyl amine, 1-propanamine, n-methyl, methyl-n-propylamine, methylpropylamine, unii-vw1h32h1qm, n-propylmethylamine, n-methyl-1-propylamine, vw1h32h1qm
N-methylpropan-1-amine

Specifications

627-35-0
62°C to 64°C
Amine-like
MFCD00009361
UN2733
GVWISOJSERXQBM-UHFFFAOYSA-N
N-methylpropan-1-amine
12315
97%
0.713
−1.1°C (30°F)
C4H11N
1 g
n-methylpropylamine, n-methyl-n-propylamine, methyl propyl amine, 1-propanamine, n-methyl, methyl-n-propylamine, methylpropylamine, unii-vw1h32h1qm, n-propylmethylamine, n-methyl-1-propylamine, vw1h32h1qm
CCCNC
73.139
73.14
N-Methyl-1-propylamine
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H225-H314-H318
Highly flammable liquid and vapour.
Causes severe skin burns and eye damage.
Causes serious eye damage.

P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c

H225-H302+H312+H332-H314-H335

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: AMINES, FLAMMABLE, CORROSIVE, N.O.S.

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only