missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N-Methyl-2-(morpholinosulfonyl)benzylamine, 97%, Thermo Scientific™

Chemical Identifiers

CAS 941717-08-4
Molecular Formula C12H18N2O3S
Molecular Weight (g/mol) 270.35
MDL Number MFCD09879987
InChI Key ASGSHKVYZRSASF-UHFFFAOYSA-N
Synonym n-methyl-2-morpholinosulfonyl benzylamine, 4-2-methylamino methyl phenylsulphonyl morpholine, methyl 2-morpholine-4-sulfonyl phenyl methyl amine, 4-2-methylamino methyl phenylsulfonyl morpholine, n-methyl-1-2-morpholinosulfonyl phenyl methanamine, n-methyl-1-2-morpholine-4-sulfonyl phenyl methanamine
PubChem CID 24229774
IUPAC Name N-methyl-1-(2-morpholin-4-ylsulfonylphenyl)methanamine
SMILES CNCC1=CC=CC=C1S(=O)(=O)N1CCOCC1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
CC69646CB
SDS View Documents Promotion Details
Thermo Scientific Chemicals
CC69646CB
250mg Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC69646DA
SDS View Documents Promotion Details
Thermo Scientific Chemicals
CC69646DA
1g Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

941717-08-4
270.35
ASGSHKVYZRSASF-UHFFFAOYSA-N
24229774
CNCC1=CC=CC=C1S(=O)(=O)N1CCOCC1
C12H18N2O3S
MFCD09879987
n-methyl-2-morpholinosulfonyl benzylamine, 4-2-methylamino methyl phenylsulphonyl morpholine, methyl 2-morpholine-4-sulfonyl phenyl methyl amine, 4-2-methylamino methyl phenylsulfonyl morpholine, n-methyl-1-2-morpholinosulfonyl phenyl methanamine, n-methyl-1-2-morpholine-4-sulfonyl phenyl methanamine
N-methyl-1-(2-morpholin-4-ylsulfonylphenyl)methanamine

Specifications

941717-08-4
100.0
Amber Glass Bottle
MFCD09879987
n-methyl-2-morpholinosulfonyl benzylamine, 4-2-methylamino methyl phenylsulphonyl morpholine, methyl 2-morpholine-4-sulfonyl phenyl methyl amine, 4-2-methylamino methyl phenylsulfonyl morpholine, n-methyl-1-2-morpholinosulfonyl phenyl methanamine, n-methyl-1-2-morpholine-4-sulfonyl phenyl methanamine
CNCC1=CC=CC=C1S(=O)(=O)N1CCOCC1
270.35
270.35
N-methyl-2-(morpholinosulfonyl)benzylamine
96.5
97%
C12H18N2O3S
250mg
ASGSHKVYZRSASF-UHFFFAOYSA-N
N-methyl-1-(2-morpholin-4-ylsulfonylphenyl)methanamine
24229774
97%
Safety and Handling

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

SDS
missing translation for 'documents'

missing translation for 'documents'