missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

n-methyl-(2-thien-2-ylpyrimidin-5-yl)methylamine, 90%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals CC66546DA

 View more versions of this product

Catalog No. CC66546DA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

Specifications

Specifications

N-methyl-(2-thien-2-ylpyrimidin-5-yl)methylamine
89.5
90%
C10H11N3S
1g
VZHVJAYQGDCZAB-UHFFFAOYSA-N
N-methyl-1-(2-thiophen-2-ylpyrimidin-5-yl)methanamine
24229733
90%
921939-14-2
100.0
Amber Glass Bottle
MFCD09879951
n-methyl-2-thien-2-ylpyrimidin-5-yl methylamine, methyl 2-thiophen-2-yl pyrimidin-5-yl methyl amine, methyl 2-2-thienyl pyrimidin-5-yl methyl amine, n-methyl-1-2-thiophen-2-yl pyrimidin-5-yl methanamine, n-methyl-2-thiophen-2-ylpyrimidin-5-yl methylamine
CNCC1=CN=C(N=C1)C2=CC=CS2
205.279
205.28

Chemical Identifiers

921939-14-2
205.279
VZHVJAYQGDCZAB-UHFFFAOYSA-N
24229733
CNCC1=CN=C(N=C1)C2=CC=CS2
C10H11N3S
MFCD09879951
n-methyl-2-thien-2-ylpyrimidin-5-yl methylamine, methyl 2-thiophen-2-yl pyrimidin-5-yl methyl amine, methyl 2-2-thienyl pyrimidin-5-yl methyl amine, n-methyl-1-2-thiophen-2-yl pyrimidin-5-yl methanamine, n-methyl-2-thiophen-2-ylpyrimidin-5-yl methylamine
N-methyl-1-(2-thiophen-2-ylpyrimidin-5-yl)methanamine
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Harmful if swallowed.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

SDS Documents

Documents

Promotions

Promotions