Learn More
n-methyl-(3-methoxythien-2-yl)methylamine, 97%, Thermo Scientific™
Chemical Identifiers
| CAS | 944450-93-5 |
|---|---|
| Molecular Formula | C7H11NOS |
| Molecular Weight (g/mol) | 157.23 |
| MDL Number | MFCD11841072 |
| InChI Key | HDALHOAZSOWZLX-UHFFFAOYSA-N |
| Synonym | 3-methoxythiophen-2-yl methyl methyl amine, 3-methoxythiophen-2-yl-n-methylmethylamine, n-methyl-3-methoxythien-2-yl methylamine, 3-methoxy-2-methylamino methyl thiophene, 1-3-methoxythiophen-2-yl-n-methylmethanamine, 3-methoxy 2-thienyl methyl methylamine |
| PubChem CID | 43811050 |
| IUPAC Name | 1-(3-methoxythiophen-2-yl)-N-methylmethanamine |
| SMILES | CNCC1=C(OC)C=CS1 |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
CC68646CB
|
Thermo Scientific Chemicals
CC68646CB |
250mg | Amber Glass Bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
CC68646DA
|
Thermo Scientific Chemicals
CC68646DA |
1g | Amber Glass Bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
| 944450-93-5 | |
| 157.23 | |
| HDALHOAZSOWZLX-UHFFFAOYSA-N | |
| 43811050 | |
| CNCC1=C(OC)C=CS1 |
| C7H11NOS | |
| MFCD11841072 | |
| 3-methoxythiophen-2-yl methyl methyl amine, 3-methoxythiophen-2-yl-n-methylmethylamine, n-methyl-3-methoxythien-2-yl methylamine, 3-methoxy-2-methylamino methyl thiophene, 1-3-methoxythiophen-2-yl-n-methylmethanamine, 3-methoxy 2-thienyl methyl methylamine | |
| 1-(3-methoxythiophen-2-yl)-N-methylmethanamine |
Specifications
| 944450-93-5 | |
| 100.0 | |
| Amber Glass Bottle | |
| MFCD11841072 | |
| 3-methoxythiophen-2-yl methyl methyl amine, 3-methoxythiophen-2-yl-n-methylmethylamine, n-methyl-3-methoxythien-2-yl methylamine, 3-methoxy-2-methylamino methyl thiophene, 1-3-methoxythiophen-2-yl-n-methylmethanamine, 3-methoxy 2-thienyl methyl methylamine | |
| CNCC1=C(OC)C=CS1 | |
| 157.23 | |
| 157.24 | |
| N-methyl-(3-methoxythien-2-yl)methylamine |
| 96.5 | |
| 97% | |
| C7H11NOS | |
| 250mg | |
| HDALHOAZSOWZLX-UHFFFAOYSA-N | |
| 1-(3-methoxythiophen-2-yl)-N-methylmethanamine | |
| 43811050 | |
| 97% |
Safety and Handling
GHS H Statement:
Harmful if swallowed.
Causes serious eye damage.
Causes severe skin burns and eye damage.
GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger