missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N-Methyl-3-methylbenzamide, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals H56299MD

 View more versions of this product

Catalog No. AAH56299MD


Explore available promotions

Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

N-Methyl-3-methylbenzamide
C9H11NO
250 mg
WKSODUAHKWBHQZ-UHFFFAOYSA-N
N,3-dimethylbenzamide
966002
97%
74786-81-5
MFCD01211937
n-methyl-3-methylbenzamide, 3,n-dimethylbenzamide, benzamide,n,3-dimethyl, benzamide, n,3-dimethyl-9ci, n-methyl 3-methylphenyl carboxamide
CC1=CC=CC(=C1)C(=O)NC
149.193
149.18

Chemical Identifiers

74786-81-5
149.193
WKSODUAHKWBHQZ-UHFFFAOYSA-N
966002
CC1=CC=CC(=C1)C(=O)NC
C9H11NO
MFCD01211937
n-methyl-3-methylbenzamide, 3,n-dimethylbenzamide, benzamide,n,3-dimethyl, benzamide, n,3-dimethyl-9ci, n-methyl 3-methylphenyl carboxamide
N,3-dimethylbenzamide
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions