missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing
N-Methyl-4-(1H-pyrazol-1-ylmethyl)benzylamine, 97%, Thermo Scientific™
Chemical Identifiers
CAS
892502-08-8
Molecular Formula
C12H15N3
Molecular Weight (g/mol)
201.27
MDL Number
MFCD08690302
InChI Key
BQLXMVISWVMAID-UHFFFAOYSA-N
Synonym
n-methyl-4-1h-pyrazol-1-ylmethyl benzylamine, methyl 4-pyrazol-1-ylmethyl phenyl methyl amine, methyl 4-1h-pyrazole-1-yl methylbenzyl amine, methyl 4-pyrazolylmethyl phenyl methyl amine, benzenemethanamine,n-methyl-4-1h-pyrazol-1-ylmethyl, n-methyl-1-4-1h-pyrazol-1-ylmethyl phenyl methanamine, 1-4-1h-pyrazol-1-yl methyl phenyl-n-methylmethanamine, n-methyl-1-4-1h-pyrazol-1-yl methyl phenyl methanamine, 3vr
PubChem CID
18525877
IUPAC Name
N-methyl-1-[4-(pyrazol-1-ylmethyl)phenyl]methanamine
SMILES
CNCC1=CC=C(CN2C=CC=N2)C=C1
View More Specs
Products
2
Description
Specifications
Chemical Identifiers
CAS
892502-08-8
Molecular Weight (g/mol)
201.27
InChI Key
BQLXMVISWVMAID-UHFFFAOYSA-N
PubChem CID
18525877
SMILES
CNCC1=CC=C(CN2C=CC=N2)C=C1
Molecular Formula
C12H15N3
MDL Number
MFCD08690302
Synonym
n-methyl-4-1h-pyrazol-1-ylmethyl benzylamine, methyl 4-pyrazol-1-ylmethyl phenyl methyl amine, methyl 4-1h-pyrazole-1-yl methylbenzyl amine, methyl 4-pyrazolylmethyl phenyl methyl amine, benzenemethanamine,n-methyl-4-1h-pyrazol-1-ylmethyl, n-methyl-1-4-1h-pyrazol-1-ylmethyl phenyl methanamine, 1-4-1h-pyrazol-1-yl methyl phenyl-n-methylmethanamine, n-methyl-1-4-1h-pyrazol-1-yl methyl phenyl methanamine, 3vr
IUPAC Name
N-methyl-1-[4-(pyrazol-1-ylmethyl)phenyl]methanamine
Specifications
CAS
892502-08-8
CAS Max %
100.0
Packaging
Amber Glass Bottle
MDL Number
MFCD08690302
Synonym
n-methyl-4-1h-pyrazol-1-ylmethyl benzylamine, methyl 4-pyrazol-1-ylmethyl phenyl methyl amine, methyl 4-1h-pyrazole-1-yl methylbenzyl amine, methyl 4-pyrazolylmethyl phenyl methyl amine, benzenemethanamine,n-methyl-4-1h-pyrazol-1-ylmethyl, n-methyl-1-4-1h-pyrazol-1-ylmethyl phenyl methanamine, 1-4-1h-pyrazol-1-yl methyl phenyl-n-methylmethanamine, n-methyl-1-4-1h-pyrazol-1-yl methyl phenyl methanamine, 3vr
SMILES
CNCC1=CC=C(CN2C=CC=N2)C=C1
Molecular Weight (g/mol)
201.27
Formula Weight
201.27
Chemical Name or Material
N-methyl-4-(1H-pyrazol-1-ylmethyl)benzylamine
CAS Min %
96.5
Assay Percent Range
97%
Molecular Formula
C12H15N3
Quantity
250mg
InChI Key
BQLXMVISWVMAID-UHFFFAOYSA-N
IUPAC Name
N-methyl-1-[4-(pyrazol-1-ylmethyl)phenyl]methanamine
PubChem CID
18525877
Percent Purity
97%
Safety and Handling
Safety and Handling
GHS H Statement: Harmful if swallowed. Causes serious eye damage. Causes severe skin burns and eye damage.
GHS P Statement: IF SWALLOWED: rinse mouth. Do NOT induce vomiting. Wear eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician. IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
SDS
missing translation for 'documents'