missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N-Methyl-[4-(3-methyl-1,2,4-oxadiazol-5-yl)phenyl]methylamine, 95%, Thermo Scientific™

Chemical Identifiers

CAS 884507-32-8
Molecular Formula C11H13N3O
Molecular Weight (g/mol) 203.25
MDL Number MFCD09025868
InChI Key MUCOLQGVWQXQMX-UHFFFAOYSA-N
Synonym n-methyl-4-3-methyl-1,2,4-oxadiazol-5-yl phenyl methylamine, methyl 4-3-methyl-1,2,4-oxadiazol-5-yl phenyl methyl amine, n-methyl-1-4-3-methyl-1,2,4-oxadiazol-5-yl phenyl methanamine, benzenemethanamine,n-methyl-4-3-methyl-1,2,4-oxadiazol-5-yl, methyl 4-3-methyl 1,2,4-oxadiazol-5-yl phenyl methyl amine
PubChem CID 18525856
IUPAC Name N-methyl-1-[4-(3-methyl-1,2,4-oxadiazol-5-yl)phenyl]methanamine
SMILES CNCC1=CC=C(C=C1)C1=NC(C)=NO1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
CC50946CB
SDS View Documents Promotion Details
Thermo Scientific Chemicals
CC50946CB
250mg N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC50946DA
SDS View Documents Promotion Details
Thermo Scientific Chemicals
CC50946DA
1g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

884507-32-8
203.25
MUCOLQGVWQXQMX-UHFFFAOYSA-N
18525856
CNCC1=CC=C(C=C1)C1=NC(C)=NO1
C11H13N3O
MFCD09025868
n-methyl-4-3-methyl-1,2,4-oxadiazol-5-yl phenyl methylamine, methyl 4-3-methyl-1,2,4-oxadiazol-5-yl phenyl methyl amine, n-methyl-1-4-3-methyl-1,2,4-oxadiazol-5-yl phenyl methanamine, benzenemethanamine,n-methyl-4-3-methyl-1,2,4-oxadiazol-5-yl, methyl 4-3-methyl 1,2,4-oxadiazol-5-yl phenyl methyl amine
N-methyl-1-[4-(3-methyl-1,2,4-oxadiazol-5-yl)phenyl]methanamine

Specifications

884507-32-8
100.0
C11H13N3O
250mg
MUCOLQGVWQXQMX-UHFFFAOYSA-N
N-methyl-1-[4-(3-methyl-1,2,4-oxadiazol-5-yl)phenyl]methanamine
18525856
95%
94.5
95%
MFCD09025868
n-methyl-4-3-methyl-1,2,4-oxadiazol-5-yl phenyl methylamine, methyl 4-3-methyl-1,2,4-oxadiazol-5-yl phenyl methyl amine, n-methyl-1-4-3-methyl-1,2,4-oxadiazol-5-yl phenyl methanamine, benzenemethanamine,n-methyl-4-3-methyl-1,2,4-oxadiazol-5-yl, methyl 4-3-methyl 1,2,4-oxadiazol-5-yl phenyl methyl amine
CNCC1=CC=C(C=C1)C1=NC(C)=NO1
203.25
203.24
N-methyl-[4-(3-methyl-1, 2, 4-oxadiazol-5-yl)phenyl]methylamine
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS
missing translation for 'documents'

missing translation for 'documents'