missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

n-methyl-(4-phenyltetrahydropyran-4-yl)methylamine, 97%, Thermo Scientific™

Chemical Identifiers

CAS 958443-30-6
Molecular Formula C13H19NO
Molecular Weight (g/mol) 205.301
MDL Number MFCD11841073
InChI Key OHCOJCIAAGLEHJ-UHFFFAOYSA-N
Synonym n-methyl-1-4-phenyltetrahydro-2h-pyran-4-yl methanamine, methyl 4-phenyloxan-4-yl methyl amine, n-methyl-4-phenyltetrahydropyran-4-yl methylamine, n-methyl-1-4-phenyltetrahydro-2h-pyran-4-yl methylamine, 4-methylamino methyl-4-phenyltetrahydro-2h-pyran, methyl 4-phenyl 2h-3,4,5,6-tetrahydropyran-4-yl methyl amine
PubChem CID 33589539
IUPAC Name N-methyl-1-(4-phenyloxan-4-yl)methanamine
SMILES CNCC1(CCOCC1)C2=CC=CC=C2
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
CC69146CB
SDS View Documents
Thermo Scientific Chemicals
CC69146CB
250mg Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC69146DA
SDS View Documents
Thermo Scientific Chemicals
CC69146DA
1g Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

958443-30-6
205.301
OHCOJCIAAGLEHJ-UHFFFAOYSA-N
33589539
CNCC1(CCOCC1)C2=CC=CC=C2
C13H19NO
MFCD11841073
n-methyl-1-4-phenyltetrahydro-2h-pyran-4-yl methanamine, methyl 4-phenyloxan-4-yl methyl amine, n-methyl-4-phenyltetrahydropyran-4-yl methylamine, n-methyl-1-4-phenyltetrahydro-2h-pyran-4-yl methylamine, 4-methylamino methyl-4-phenyltetrahydro-2h-pyran, methyl 4-phenyl 2h-3,4,5,6-tetrahydropyran-4-yl methyl amine
N-methyl-1-(4-phenyloxan-4-yl)methanamine

Specifications

958443-30-6
100.0
Amber Glass Bottle
MFCD11841073
n-methyl-1-4-phenyltetrahydro-2h-pyran-4-yl methanamine, methyl 4-phenyloxan-4-yl methyl amine, n-methyl-4-phenyltetrahydropyran-4-yl methylamine, n-methyl-1-4-phenyltetrahydro-2h-pyran-4-yl methylamine, 4-methylamino methyl-4-phenyltetrahydro-2h-pyran, methyl 4-phenyl 2h-3,4,5,6-tetrahydropyran-4-yl methyl amine
CNCC1(CCOCC1)C2=CC=CC=C2
205.301
205.3
N-methyl-(4-phenyltetrahydropyran-4-yl)methylamine
96.5
97%
C13H19NO
250mg
OHCOJCIAAGLEHJ-UHFFFAOYSA-N
N-methyl-1-(4-phenyloxan-4-yl)methanamine
33589539
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Harmful if swallowed.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

SDS
missing translation for 'documents'

missing translation for 'documents'