missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

n-methyl-{[5-(morpholinomethyl)-2-furyl]methyl}amine, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals CC72546DA

 View more versions of this product

Catalog No. CC72546DA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

Specifications

Specifications

N-methyl-{[5-(morpholinomethyl)-2-furyl]methyl}amine
96.5
97%
C11H18N2O2
1g
RVGSMPQAXGUMNM-UHFFFAOYSA-N
N-methyl-1-[5-(morpholin-4-ylmethyl)furan-2-yl]methanamine
16495000
97%
893741-66-7
100.0
Amber Glass Bottle
MFCD06803236
n-methyl-5-morpholinomethyl-2-furyl methyl amine, methyl 5-morpholin-4-ylmethyl furan-2-yl methyl amine, n-methyl-1-5-morpholin-4-ylmethyl furan-2-yl methylamine, 2-furanmethanamine,n-methyl-5-4-morpholinylmethyl, 4-5-methylamino methyl furan-2-ylmethyl morpholine, methyl 5-morpholin-4-ylmethyl 2-furyl methyl amine, n-methyl-1-5-morpholinomethyl furan-2-yl methanamine, n-methyl-1-5-morpholin-4-yl methyl furan-2-yl methanamine
CNCC1=CC=C(CN2CCOCC2)O1
210.28
210.28

Chemical Identifiers

893741-66-7
210.28
RVGSMPQAXGUMNM-UHFFFAOYSA-N
16495000
CNCC1=CC=C(CN2CCOCC2)O1
C11H18N2O2
MFCD06803236
n-methyl-5-morpholinomethyl-2-furyl methyl amine, methyl 5-morpholin-4-ylmethyl furan-2-yl methyl amine, n-methyl-1-5-morpholin-4-ylmethyl furan-2-yl methylamine, 2-furanmethanamine,n-methyl-5-4-morpholinylmethyl, 4-5-methylamino methyl furan-2-ylmethyl morpholine, methyl 5-morpholin-4-ylmethyl 2-furyl methyl amine, n-methyl-1-5-morpholinomethyl furan-2-yl methanamine, n-methyl-1-5-morpholin-4-yl methyl furan-2-yl methanamine
N-methyl-1-[5-(morpholin-4-ylmethyl)furan-2-yl]methanamine
Safety and Handling

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.
Harmful if swallowed.
Causes serious eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

SDS Documents

Documents

Promotions

Promotions