missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
N-Methyl-D-aspartic acid, 98%, Thermo Scientific Chemicals
$145.76 - $556.62
Chemical Identifiers
CAS | 6384-92-5 |
---|---|
Molecular Formula | C5H9NO4 |
Molecular Weight (g/mol) | 147.1 |
MDL Number | MFCD00004226 |
InChI Key | HOKKHZGPKSLGJE-GSVOUGTGSA-N |
Synonym | n-methyl-d-aspartic acid, nmda, n-methyl-d-aspartate, n-methylaspartate, d-aspartic acid, n-methyl, methyl aspartic acid, n-me-d-asp-oh, r-2-methylamino succinic acid, unii-1903b9q6pi, n methyl d aspartate |
PubChem CID | 22880 |
ChEBI | CHEBI:31882 |
IUPAC Name | (2R)-2-(methylamino)butanedioic acid |
SMILES | CNC(CC(=O)O)C(=O)O |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | |||||
AC329190500
|
Thermo Scientific Chemicals
329190500 |
50 mg | Glass bottle |
Each for $145.76
|
|
|||||
AC329192500
|
Thermo Scientific Chemicals
329192500 |
250 mg | Glass bottle |
Each for $556.62
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
6384-92-5 | |
147.1 | |
HOKKHZGPKSLGJE-GSVOUGTGSA-N | |
22880 | |
(2R)-2-(methylamino)butanedioic acid |
C5H9NO4 | |
MFCD00004226 | |
n-methyl-d-aspartic acid, nmda, n-methyl-d-aspartate, n-methylaspartate, d-aspartic acid, n-methyl, methyl aspartic acid, n-me-d-asp-oh, r-2-methylamino succinic acid, unii-1903b9q6pi, n methyl d aspartate | |
CHEBI:31882 | |
CNC(CC(=O)O)C(=O)O |
Specifications
6384-92-5 | |
100.0 | |
White | |
98% | |
C5H9NO4 | |
50 mg | |
− 28.00 (20.00°C c=1,HCl 6N) | |
n-methyl-d-aspartic acid, nmda, n-methyl-d-aspartate, n-methylaspartate, d-aspartic acid, n-methyl, methyl aspartic acid, n-me-d-asp-oh, r-2-methylamino succinic acid, unii-1903b9q6pi, n methyl d aspartate | |
HOKKHZGPKSLGJE-GSVOUGTGSA-N | |
(2R)-2-(methylamino)butanedioic acid | |
22880 | |
147.1 | |
Powder |
97.5 | |
187°C to 192°C | |
Authentic | |
Glass bottle | |
MFCD00004226 | |
15, 6749 | |
-28 | |
Solubility in water: slightly soluble in water. Other solubilities: soluble in dilute aqueous base,slightly soluble i | |
CNC(CC(=O)O)C(=O)O | |
147.1 | |
CHEBI:31882 | |
98% | |
N-Methyl-D-aspartic acid, 98% |
Safety and Handling
LIGHT SENSITIVE
RTECSNumber : CI9457000
RUO – Research Use Only