missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

n-methyl-n-(3-piperidin-1-ylbenzyl)amine, 90%, Thermo Scientific™

Chemical Identifiers

CAS 859850-65-0
Molecular Formula C13H20N2
Molecular Weight (g/mol) 204.317
MDL Number MFCD07772817
InChI Key WHELHMLNRPDTJU-UHFFFAOYSA-N
Synonym n-methyl-n-3-piperidin-1-ylbenzyl amine, methyl 3-piperidin-1-yl phenyl methyl amine, methyl 3-piperidylphenyl methyl amine, benzenemethanamine,n-methyl-3-1-piperidinyl, n-methyl-1-3-piperidin-1-ylphenyl methanamine, n-methyl-1-3-piperidin-1-yl phenyl methanamine
PubChem CID 7162073
IUPAC Name N-methyl-1-(3-piperidin-1-ylphenyl)methanamine
SMILES CNCC1=CC(=CC=C1)N2CCCCC2
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
CC34046CB
SDS View Documents
Thermo Scientific Chemicals
CC34046CB
250mg Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC34046DA
SDS View Documents
Thermo Scientific Chemicals
CC34046DA
1g Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

859850-65-0
204.317
WHELHMLNRPDTJU-UHFFFAOYSA-N
7162073
CNCC1=CC(=CC=C1)N2CCCCC2
C13H20N2
MFCD07772817
n-methyl-n-3-piperidin-1-ylbenzyl amine, methyl 3-piperidin-1-yl phenyl methyl amine, methyl 3-piperidylphenyl methyl amine, benzenemethanamine,n-methyl-3-1-piperidinyl, n-methyl-1-3-piperidin-1-ylphenyl methanamine, n-methyl-1-3-piperidin-1-yl phenyl methanamine
N-methyl-1-(3-piperidin-1-ylphenyl)methanamine

Specifications

859850-65-0
100.0
Amber Glass Bottle
MFCD07772817
n-methyl-n-3-piperidin-1-ylbenzyl amine, methyl 3-piperidin-1-yl phenyl methyl amine, methyl 3-piperidylphenyl methyl amine, benzenemethanamine,n-methyl-3-1-piperidinyl, n-methyl-1-3-piperidin-1-ylphenyl methanamine, n-methyl-1-3-piperidin-1-yl phenyl methanamine
CNCC1=CC(=CC=C1)N2CCCCC2
204.317
204.32
N-methyl-N-(3-piperidin-1-ylbenzyl)amine
89.5
95%
C13H20N2
250mg
WHELHMLNRPDTJU-UHFFFAOYSA-N
N-methyl-1-(3-piperidin-1-ylphenyl)methanamine
7162073
95%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Causes serious eye damage.
Causes severe skin burns and eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

SDS
missing translation for 'documents'

missing translation for 'documents'