missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Methyl-N-(trifluoromethylthio)aniline 98.0+%, TCI America™
$251.65 - $892.89
Chemical Identifiers
| CAS | 66476-44-6 |
|---|---|
| Molecular Formula | C8H8F3NS |
| Molecular Weight (g/mol) | 207.214 |
| InChI Key | TYFGNILTFVMYGX-UHFFFAOYSA-N |
| PubChem CID | 25013449 |
| IUPAC Name | N-methyl-N-(trifluoromethylsulfanyl)aniline |
| SMILES | CN(C1=CC=CC=C1)SC(F)(F)F |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
M2595200MG
|
TCI America
M2595200MG |
200 mg |
Each for $251.65
|
|
|||||
|
M25951G
|
TCI America
M25951G |
1 g |
Each for $892.89
|
|
|||||
Chemical Identifiers
| 66476-44-6 | |
| 207.214 | |
| 25013449 | |
| CN(C1=CC=CC=C1)SC(F)(F)F |
| C8H8F3NS | |
| TYFGNILTFVMYGX-UHFFFAOYSA-N | |
| N-methyl-N-(trifluoromethylsulfanyl)aniline |
Specifications
| 66476-44-6 | |
| C8H8F3NS | |
| 1993 | |
| CN(C1=CC=CC=C1)SC(F)(F)F | |
| 207.214 | |
| 207.21 | |
| Liquid |
| Yellow | |
| 200 mg | |
| TYFGNILTFVMYGX-UHFFFAOYSA-N | |
| N-methyl-N-(trifluoromethylsulfanyl)aniline | |
| 25013449 | |
| ≥98.0% (GC) | |
| N-Methyl-N-(trifluoromethylthio)aniline |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator