missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N-Methylaniline 98.0+%, TCI America™

Supplier:  TCI America M0147500G

 View more versions of this product

Catalog No. M0147500G


Explore available promotions

Description

Description

Specifications

Specifications

N-Methylaniline
-57°C
196°C
MFCD00008283
2294
AFBPFSWMIHJQDM-UHFFFAOYSA-N
N-methylaniline
7515
107.16
Liquid
100-61-8
Yellow
C7H9N
500 g
methylaniline, monomethylaniline, n-methyl aniline, benzenamine, n-methyl, methylphenylamine, n-methylbenzenamine, n-monomethylaniline, n-methylaminobenzene, methylamino benzene, anilinomethane
CNC1=CC=CC=C1
107.16
CHEBI:15733
≥98.0% (GC,T)

Chemical Identifiers

100-61-8
107.16
AFBPFSWMIHJQDM-UHFFFAOYSA-N
7515
N-methylaniline
C7H9N
MFCD00008283
methylaniline, monomethylaniline, n-methyl aniline, benzenamine, n-methyl, methylphenylamine, n-methylbenzenamine, n-monomethylaniline, n-methylaminobenzene, methylamino benzene, anilinomethane
CHEBI:15733
CNC1=CC=CC=C1
Videos
Safety and Handling

Safety and Handling

EINECSNumber : (3)-0106

RTECSNumber : BY4550000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions