Learn More
N-Methylbenzylamine, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 106045000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
N-Methylbenzylamine | |
103-67-3 | |
100.0 | |
Colorless to Yellow | |
184°C to 189°C | |
Authentic | |
Glass bottle | |
1.5214 to 1.5234 | |
MFCD00008289 | |
12, 1019 | |
n-methylbenzylamine, methylbenzylamine, n-benzylmethylamine, benzyl methyl amine, benzenemethanamine, n-methyl, benzylmethylamine, n-methy-n-benzylamine, n-benzyl-n-methylamine, benzylamine, n-methyl, n-methyl phenyl methanamine | |
RIWRFSMVIUAEBX-UHFFFAOYSA-N | |
N-methyl-1-phenylmethanamine | |
7669 | |
97% |
97% | |
96.0 | |
-24°C | |
0.9400g/mL | |
63°C | |
97% | |
C8H11N | |
C6H5CH2NHCH3 | |
500 mL | |
0.94 | |
Solubility in water: 65g/L (20°C) | |
CNCC1=CC=CC=C1 | |
121.18 | |
121.18 | |
Liquid |
Chemical Identifiers
103-67-3 | |
121.18 | |
RIWRFSMVIUAEBX-UHFFFAOYSA-N | |
7669 | |
CNCC1=CC=CC=C1 |
C8H11N | |
MFCD00008289 | |
n-methylbenzylamine, methylbenzylamine, n-benzylmethylamine, benzyl methyl amine, benzenemethanamine, n-methyl, benzylmethylamine, n-methy-n-benzylamine, n-benzyl-n-methylamine, benzylamine, n-methyl, n-methyl phenyl methanamine | |
N-methyl-1-phenylmethanamine |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
Toxic if swallowed.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for
GHS Signal Word: Danger
EINECSNumber : 203-133-4
TSCA : TSCA
RUO – Research Use Only