missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N-Methylcaprolactam, 96%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1777622

 View more versions of this product

Catalog No. AAA1777622


Explore available promotions

Description

Description

N-Methylcaprolactam was used to study the inhibition mechanism of poly-[N-vinylcaprolactam] (PVCAP) and its related compounds against hydrate formation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Methylcaprolactam was used to study the inhibition mechanism of poly-[N-vinylcaprolactam] (PVCAP) and its related compounds against hydrate formation.

Solubility
Fully miscible with water.

Notes
Hygroscopic, store away from moisture. Store in a tightly closed container in a cool, dry, well-ventilated area. Store under dry inert gas.
Specifications

Specifications

N-Methylcaprolactam
0.991
103°C (217°F)
1.484
100 g
n-methylcaprolactam, 1-methylcaprolactam, n-methyl-.epsilon.-caprolactam, 2h-azepin-2-one, hexahydro-1-methyl, n-methyl-e-caprolactam, epsilon-caprolactam, n-methyl, 2h-azepin-2-one, hexahydro-n-methyl, hexahydro-n-methyl-2h-azepin-2-one, n-methyl-epsilon-caprolactam, n-methyl caprolactam
ZWXPDGCFMMFNRW-UHFFFAOYSA-N
1-methylazepan-2-one
17369
96%
2556-73-2
106°C to 108°C (6 mmHg)
C7H13NO
MFCD00003263
Hygroscopic
Fully miscible with water.
CN1CCCCCC1=O
127.187
127.18

Chemical Identifiers

2556-73-2
127.187
ZWXPDGCFMMFNRW-UHFFFAOYSA-N
17369
CN1CCCCCC1=O
C7H13NO
MFCD00003263
n-methylcaprolactam, 1-methylcaprolactam, n-methyl-.epsilon.-caprolactam, 2h-azepin-2-one, hexahydro-1-methyl, n-methyl-e-caprolactam, epsilon-caprolactam, n-methyl, 2h-azepin-2-one, hexahydro-n-methyl, hexahydro-n-methyl-2h-azepin-2-one, n-methyl-epsilon-caprolactam, n-methyl caprolactam
1-methylazepan-2-one
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H311-H315-H319-H335
Toxic if swallowed.
Toxic in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H312-H315-H319-H335

EINECSNumber : 219-874-1

RTECSNumber : CM4025000

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only